Skip to Content
Merck
All Photos(4)

Key Documents

HPA012601

Sigma-Aldrich

Anti-FABP6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fatty acid-binding protein 6, Anti-GT, Anti-Gastrotropin, Anti-I-15P, Anti-I-BABP, Anti-ILBP, Anti-Ileal lipid-binding protein, Anti-Intestinal 15 kDa protein, Anti-Intestinal bile acid-binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

AFTGKFEMESEKNYDEFMKLLGISSDVIEKARNFKIVTEVQQDGQDFTWSQHYSGGHTMTNKFTVGKESNIQTMGGKMFKATVQMEGGKLVVNFPNYHQTSEIVGDKLVEVSTIGGVTYERVSKRLA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FABP6(2172)

General description

Fatty acid binding protein 6 (FABP6) is localized in the cytosol of cells in the intestine extending from the jejunum to the ascending colon. It is a 15kDa protein belonging to the fatty acid-binding protein family (FABP family).

Immunogen

Gastrotropin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Fatty acid binding protein 6 (FABP6) is involved in binding and transporting bile acids. It is a part of the farnesoid X receptor (FXR) transcription pathway which responds to bile acids and regulates their reabsorption across the ileum. It also enhances the transcriptional activity of FXR. Bile acids that are secreted in the small intestine are reabsorbed in the ileum. They activate the nuclear FXR and this in turn stimulates the expression of FABP6. The interaction between FXR and FABP6 causes a local increase in bile acid concentration around the ligand-binding domain (LBD) of FXR. This aids the delivery of ligands to the LBD.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71278

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ileal bile acid-binding protein, functionally associated with the farnesoid X receptor or the ileal bile acid transporter, regulates bile acid activity in the small intestine.
M Nakahara
The Journal of Biological Chemistry, 280(51), 42283-42289 (2005)
M Fujita et al.
European journal of biochemistry, 233(2), 406-413 (1995-10-15)
We have isolated a cDNA encoding a human intestinal 15-kDa protein (I-15P) from a human ileal lambda gt 11 cDNA library, using a full-length rat I-15P cDNA. One clone encompassed 571 nucleotides and encoded a 128-amino-acid protein with a calculated
Changming Fang et al.
Cancer research, 67(19), 9039-9046 (2007-10-03)
Ileal bile acid binding protein (IBABP) is the only cytosolic protein known to bind and transport bile acids. Because IBABP is reportedly up-regulated in colorectal cancer, it has been suggested as a link between bile acids and the risk of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service