Skip to Content
Merck
All Photos(6)

Key Documents

HPA012128

Sigma-Aldrich

Anti-TNIK antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TRAF2 and NCK-interacting protein kinase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

QGPALTASQSVHEQPTKGLSGFQEALNVTSHRVEMPRQNSDPTSENPPLPTRIEKFDRSSWLRQEEDIPPKVPQRTTSISPALARKNSPGNGSALGPRLGSQPIRASNPDLRRTEPILESPLQRTSSGSSSSSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNIK(23043)

Immunogen

TRAF2 and NCK-interacting protein kinase recombinant protein epitope signature tag (PrEST)

Application

Anti-TNIK antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

TNIK (TRAF2 and NCK interacting kinase) is a protein kinase belonging to the germinal center kinase family. It consists of an N-terminal kinase domain. It is localized in the nucleus. TNIK is highly involved in the cytoskeleton assembly and neuronal dendrite extension. In cytoskeleton arrangement, it directly interacts with Rap2 for the proper distribution and spreading of cells. It also stimulates c-Jun N-terminal kinase for the cytoskeleton regulation. TNIK acts as transcription activator during Wnt signaling, which is an essetial step for several physiological processes, including cell proliferation, differentiation, motility, apoptosis. It also plays an important role in the intestinal crypts and colorectal cancer cells. It has been suggested that TNIK can be a potential pharmacological kinase inhibitor for the treatment of colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71436

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hidenori Takahashi et al.
BMC cancer, 15, 794-794 (2015-10-27)
The potential of expression profiling using microarray analysis as a tool to predict the prognosis for different types of cancer has been realized. This study aimed to identify a novel biomarker for colorectal cancer (CRC). The expression profiles of cancer
C A Fu et al.
The Journal of biological chemistry, 274(43), 30729-30737 (1999-10-16)
Germinal center kinases (GCKs) compose a subgroup of the Ste20 family of kinases. Here we describe the cloning and characterization of a novel GCK family kinase, Traf2- and Nck-interacting kinase (TNIK) that interacts with both Traf2 and Nck. TNIK encodes
D-H Yu et al.
Oncogenesis, 2, e89-e89 (2014-02-26)
Traf2- and Nck-interacting kinase (TNIK) is one of the germinal center kinase family members involved in cytoskeleton organization and neuronal dendrite extension. Emerging evidence supports that TNIK is essential for activation of WNT signaling pathway in colon cancer growth. To
Alain C Burette et al.
The Journal of comparative neurology, 523(13), 1913-1924 (2015-03-11)
Tumor necrosis factor receptor-associated factor 2 (TRAF2)- and noncatalytic region of tyrosine kinase (NCK)-interacting kinase (TNIK) has been identified as an interactor in the psychiatric risk factor, Disrupted in Schizophrenia 1 (DISC1). As a step toward deciphering its function in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service