Skip to Content
Merck
All Photos(5)

Key Documents

HPA011851

Sigma-Aldrich

Anti-PTPRR antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Ch-1PTPase, Anti-NC-PTPCOM1, Anti-Protein-tyrosine phosphatase PCPTP1, Anti-Receptor-type tyrosine-protein phosphatase R precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

YDPSLNLLAMDGQDLEVENLPIPAANVIVVTLQMDVNKLNITLLRIFRQGVAAALGLLPQQVHINRLIGKKNSIELFVSPINRKTGISDALPSEEVLRSLNINVLHQSLSQFGITEVSPEKNVLQGQHE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTPRR(5801)

Immunogen

Receptor-type tyrosine-protein phosphatase R precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PTPRR antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PTPRR (Protein tyrosine phosphatase, receptor type, R) is a transmembrane protein-tyrosine phosphatase (PTP) belonging to the larger family of PTPs. It is expressed in the brain uterus, placenta, and lower gastrointestinal tract. It is associated with the cell proliferation and differentiation. It plays a vital role in embryonic development and in adult physiology. PTPRR negatively regulates the extracellular regulated kinase (ERK) signaling pathway. It is associated with a common, chronic, and recurrent mental disease, major depressive disorder (MDD). It also participates in cell metastasis through MAPK signaling pathways in the invasive cervical cancer, which proves PTPRR as a potential biomarker of invasive cervical cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72115

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Woźniak et al.
Histology and histopathology, 29(5), 635-639 (2013-11-19)
Two classes of proteins, namely tyrosine kinases (PTK) and phosphatases (PTP), play an important role in cell proliferation and differentiation, thus leading to an acceleration or inhibition of tumour growth. The role of the above proteins in colorectal carcinoma (CRC)
Cuijuan Shi et al.
Journal of affective disorders, 136(3), 591-598 (2011-11-22)
Major depressive disorder (MDD) is a common, chronic, and recurrent mental disease affecting millions of individuals worldwide. The precise mechanism by which the illness is developed remains unknown, but it has been accepted that a genetic component is very likely
K A Augustine et al.
The Anatomical record, 258(3), 221-234 (2000-03-08)
Protein tyrosine phosphatases (PTPs) play important roles in modulating signals transduced by tyrosine kinases. Certain phosphatases have been implicated as having important roles in embryonic development as well as in adult physiology. Although both kinases and phosphatases are equally important
P-H Su et al.
Oncogene, 32(1), 15-26 (2012-02-15)
Epigenetic modifications are a driving force in carcinogenesis. However, their role in cancer metastasis remains poorly understood. The present study investigated the role of DNA methylation in the cervical cancer metastasis. Here, we report evidence of the overexpression of DNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service