Skip to Content
Merck
All Photos(6)

Key Documents

HPA011765

Sigma-Aldrich

Anti-SENP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Sentrin-specific protease 1 antibody produced in rabbit, Anti-Sentrin/SUMO-specific protease SENP1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Estimated to ship on26 April 2025



Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Estimated to ship on26 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GDLRTFGQSANGQWRNSTPSSSSSLQKSRNSRSLYLETRKTSSGLSNSFAGKSNHHCHVSAYEKSFPIKPVPSPSWSGSCRRSLLSPKKTQRRHVSTAEETVQEEEREIYRQLLQMVTGK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SENP1(29843)

Related Categories

Immunogen

Sentrin-specific protease 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-SENP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SENP1 (SUMO1/sentrin specific peptidase 1) is a sentrin/SUMO-specific protease (SENPs) belonging to the de-SUMOylation protease family. It essentially regulates the androgen receptor-dependent transcription and hypoxia signaling. It exerts pro-oncogenic characteristics in various types of malignancies. Studies show that SENP1 is overexpressed in both the cancers, precancerous prostate intraepithelial neoplasia (PIN) lesions and prostate cancer tissues; however, it is correlated with the intensity and recurrence of prostate cancer. SENP1 regulates the expression of two critical bone remodeling proteins, matrix metalloproteinase 2 (MMP2) and MMP9 via HIF1a signaling pathway. Research shows the correlation between SENP1 and the metastasis in prostate cancer, since; it has identifying property as a prognostic marker.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70779

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Wang et al.
Osteoarthritis and cartilage, 24(6), 1113-1124 (2016-01-31)
To explore the hypoxic regulation of sumoylation pathways and cell viability in nucleus pulposus (NP) and annulus fibrosus (AF) cells. Expression of small ubiquitin-like modifier (SUMO) molecules, SUMO E1 activating enzymes SAE1 and SAE2, SUMO E2 conjugating enzyme UBC9, and
Martijn van Hagen et al.
Nucleic acids research, 38(6), 1922-1931 (2009-12-23)
Hypoxia-inducible factors (HIFs) are critical transcription factors that mediate cell survival during reduced oxygen conditions (hypoxia). At regular oxygen conditions (normoxia), HIF-1alpha and HIF-2alpha are continuously synthesized in cells and degraded via the ubiquitin-proteasome pathway. During hypoxia, these proteins are
Xue-Qing Wan et al.
International journal of molecular medicine, 43(1), 371-381 (2018-11-06)
Post‑translational modification via small ubiquitin‑like modifier (SUMO) is involved in the regulation of various important cellular processes. SUMO modification can be regulated at the level of conjugation, and can also be reversed by the SUMO‑specific proteases (SENPs). However, current studies
Chenchao Ma et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(12), 12729-12735 (2014-09-14)
SUMOylation is a dynamic process which can be reversed by a family of sentrin/SUMO-specific protease (SENPs). Recently, SENP1, a member of SENPs family was shown to have a pro-oncogenic role in many types of cancer. Here, we showed that SENP1
Q Wang et al.
Oncogene, 32(19), 2493-2498 (2012-06-27)
SUMO-specific protease 1 (SENP1) is a member of de-SUMOylation protease family and has an important role in the regulation of androgen receptor-dependent transcription and hypoxia signaling. This activity profile of SENP1 prompted us to investigate whether SENP1 is involved in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service