Skip to Content
Merck
All Photos(3)

Key Documents

HPA011739

Sigma-Aldrich

Anti-PARP4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-193 kDa vault protein, Anti-PARP-4, Anti-PH5P, Anti-Poly recombinant protein, Anti-VPARP, Anti-Vault poly(ADP-ribose

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ALLLEEVMNSSTLSQEVSDLVEMIWAEALGHLEHMLLKPVNRISLNDVSKAEGILLLVKAALKNGETAEQLQKMMTEFYRLIPHKGTMPKEVNLGLLAKKADLCQLIRDMVNVCETNLSKPNPPSLAKYRALRCKIEHVEQNTEEF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PARP4(143)

General description

PARP4 (Poly ADP-ribose polymerase family, member 4) is a 193kDa nuclear, NAD-dependent, vault protein encoding an ADP-ribosyltransferase-like protein. It is a subunit of ribonucleoprotein (RNP) complex belonging to the inter-α-inhibitor family. Structurally, it is composed of a catalytic domain similar to the nuclear-localized ADP-ribosyltransferase proteins (Adprt), two Adprt-like proteins (Adprtl2 and Adprtl3), and the telomere-associated protein tankyrase.

Immunogen

Poly recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PARP4 (Poly ADP-ribose polymerase family, member 4) is associated with various cellular processes including genomic DNA integrity related activities, DNA repair, replication, transcription, and apoptosis. In DNA assembly, it crucially identifies the DNA nick sites and rejoins the DNA strands. It poly(ADP-ribosyl)ates the nuclear proteins in response to any DNA damage.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70205

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V A Kickhoefer et al.
The Journal of cell biology, 146(5), 917-928 (1999-09-09)
Mammalian vaults are ribonucleoprotein (RNP) complexes, composed of a small ribonucleic acid and three proteins of 100, 193, and 240 kD in size. The 100-kD major vault protein (MVP) accounts for >70% of the particle mass. We have identified the
I H Still et al.
Genomics, 62(3), 533-536 (2000-01-25)
Poly(ADP-ribosyl)ation of nuclear proteins plays a significant role in the maintenance of genomic DNA stability. To date, four poly(ADP-ribosyl)ating proteins have been identified in humans. We now report the full-length sequence, expression profile, and chromosomal localization of a novel gene
L Jean et al.
FEBS letters, 446(1), 6-8 (1999-04-01)
Poly(ADP-ribose)polymerase is a nuclear NAD-dependent enzyme and an essential nick sensor involved in cellular processes where nicking and rejoining of DNA strands are required. The inter-alpha-inhibitor family is comprized of several plasma proteins that all harbor one or more so-called
Véronique J Bouchard et al.
Experimental hematology, 31(6), 446-454 (2003-06-28)
Poly(ADP-ribose) polymerase-1 (PARP-1) plays a primary role in the process of poly(ADP-ribosyl)ation. This posttranslational modification of nuclear proteins is activated in response to DNA damage. Having been studied for more than 30 years, PARP-1 is now known to be implicated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service