Skip to Content
Merck
All Photos(7)

Key Documents

HPA011387

Sigma-Aldrich

Anti-SGK2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Serine/threonine-protein kinase Sgk2, Anti-Serum/glucocorticoid-regulated kinase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

EVLRKEPYDRAVDWWCLGAVLYEMLHGLPPFYSQDVSQMYENILHQPLQIPGGRTVAACDLLQSLLHKDQRQRLGSKADFLEIKNHVFFSPINWDDLYHKRLTPPFNPNVTGPADLKHFDPEFTQEAVSKSIGCTPD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGK2(10110)

Immunogen

Serine/threonine-protein kinase Sgk2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SGK2 (Serum/glucocorticoid regulated kinase 2) plays a critical role in the PXR (pregnane X receptor)-regulated glucose 6-phosphatase (G6Pase) induction and gluconeogenesis. It controls the biosynthesis of glucose through the insulin-independent signal pathway. Similar to SGK1, it also triggers the renal epithelial Na+ channel (ENaC) activity. It is present downstream of phosphoinositide-3-kinase (PI3Kinase) and controls the tumor-promoting gene signaling pathways such as RAS/RAF/ERK signaling. It also reflects a stimulating activity of neutral amino acid transporter SLC6A19 (B(0)AT1) for the amino acid transport in the kidney and intestine.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70466

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maressa A Bruhn et al.
Growth factors (Chur, Switzerland), 28(6), 394-408 (2010-10-06)
The serum and glucocorticoid kinase (SGK) family of serine/threonine kinases consists of three isoforms, SGK-1, SGK-2 and SGK-3. This family of kinases is highly homologous to the AKT kinase family, sharing similar upstream activators and downstream targets. SGKs have been
B Friedrich et al.
Pflugers Archiv : European journal of physiology, 445(6), 693-696 (2003-03-13)
The serum- and glucocorticoid-inducible kinase 1 (SGK1) has been identified as a signalling molecule up-regulated by aldosterone, which stimulates the renal epithelial Na(+) channel ENaC. It is therefore thought to participate in the antinatriuretic action of this hormone. More recently
Christoph Böhmer et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 25(6), 723-732 (2010-06-01)
The neutral amino acid transporter SLC6A19 (B(0)AT1) plays a decisive role in transport of neutral amino acids in the kidney and intestine. Recently, mutations in SLC6A19 were identified that result in severe neutral aminoaciduria known as Hartnup disorder. SLC6A19 expression
Saki Gotoh et al.
The Journal of pharmacology and experimental therapeutics, 348(1), 131-140 (2013-11-10)
Drug activation of the human nuclear pregnane X receptor (PXR) induced gluconeogenic genes and increased glucose production. In this study, we have determined that serum- and glucocorticoid-regulated kinase 2 (SGK2) is an essential factor that mediates this PXR-regulated glucose 6-phosphatase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service