Skip to Content
Merck
All Photos(6)

Key Documents

HPA011337

Sigma-Aldrich

Anti-CASP6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Apoptotic protease Mch-2, Anti-CASP-6, Anti-Caspase-6 precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DVPVIPLDVVDNQTEKLDTNITEVDAASVYTLPAGADFLMCYSVAEGYYSHRETVNGSWYIQDLCEMLGKYGSSLEFTELLTLVNRKVSQRRVDFCKDPSAIGKKQV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CASP6(839)

General description

CASP6 (caspase 6) belongs to the family of cysteine proteases called caspases. Caspases can be divided into inflammatory capases, apoptosis initiators and effector caspases, and CASP6 belongs to the effector class. It exists as a dimeric zymogen. Each CASP6 monomer contains a short pro-domain called CARD (caspase-recruitment domain) or DED (death effector domain), a large subunit (p20) and a small subunit (p10), intervened by an intersubunit linker (L). It is expressed in both fetal and adult human tissues, though it has a higher level of expression during development, and decreased levels in adult tissues. In fetus, CASP6 has the highest level of expression in the gastrointestinal tract and the lowest in brain. In adults, it is predominantly expressed in colon, lung, stomach, kidney and liver.

Immunogen

Caspase-6 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CASP6 (caspase 6) plays a key role in apoptosis, and cleaves proteins which contain (V/I/T/L)E(G/D)ID sites. It has two classes of substrate proteins- proteins involved in nuclear structure or function and intermediate filament proteins. Lamin A, B and C, DNA topoisomerase I, CBP/p300, nuclear death domain protein p84N5, nuclear matrix protein SATB1, emerin, NuMA, DFF40, and PARP etc. act as its substrates in nucleus. Chromatin condensation in apoptosis is a result of proteolysis of lamin A by CASP6. In cytoplasm, it acts upon desmin, vimentin and cytokeratin, thereby modulating cell structure and function. It is responsible for the degradation of axons in the primary cortex. It enhances the synthesis of amyloid β peptide (Aβ), and is highly expressed in the hippocampus and cortex in the brains of Alzheimer′s disease (AD) patients. CASP6 is also involved in the impairment of cognitive abilities in AD patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70822

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Zhao et al.
Cellular and molecular neurobiology, 34(3), 369-378 (2013-12-24)
Tau truncation is widely detected in Alzheimer's disease brain. Caspases activation is suggested to play a significant role in tau truncation at Aspartate 421 (D421) according to their ability to cleave recombinant tau in vitro. Ample evidence has shown that
Qin Cao et al.
Acta crystallographica. Section D, Biological crystallography, 70(Pt 1), 58-67 (2014-01-15)
Caspase 6 (CASP6) is a neuron degeneration-related protease and is widely considered to be a potential drug-design target against neurodegenerative diseases such as Huntington's disease and Alzheimer's disease. The N-terminal pro-peptide of CASP6, also referred to as the pro-domain, contains
Jasmine Ramcharitar et al.
Neurobiology of aging, 34(7), 1815-1824 (2013-02-14)
Caspase-6 (Casp6), a cysteinyl protease that induces axonal degeneration, is activated early in Alzheimer Disease (AD) brains. To determine whether Casp6 activation is responsible for early cognitive impairment, we investigated the abundance of Casp6 activity, paired helical filament-1 (PHF-1) phosphorylated
Nelly Godefroy et al.
PloS one, 8(11), e79313-e79313 (2013-11-23)
Caspase-6 is an effector caspase that has not been investigated thoroughly despite the fact that Caspase-6 is strongly activated in Alzheimer disease brains. To understand the full physiological impact of Caspase-6 in humans, we investigated Caspase-6 expression. We performed western

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service