Skip to Content
Merck
All Photos(4)

Key Documents

HPA011090

Sigma-Aldrich

Anti-SEMA4C antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Semaphorin-4C precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VDGELYSATLNNFLGTEPIILRNMGPHHSMKTEYLAFWLNEPHFVGSAYVPESVGSFTGDDDKVYFFFRERAVESDCYAEQVVARVARVCKGDMGGARTLQRKWTTFLKARLACSAPNWQLYF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SEMA4C(54910)

General description

SEMA4C (semaphorin 4C) is a transmembrane neural protein which belongs to the family of semaphorin proteins. It has a cytoplasmic domain made of 146 amino acids, which has a proline rich region. The C-terminal contains a PDZ binding domain. This protein is expressed in neurons of embryos, and predominantly in postnatal and adult brain.

Immunogen

Semaphorin-4C precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The function of SEMA4C (semaphorin 4C) is not well defined. It functions as a ligand for cell-sruface signaling by interacting with SEMCAP-1 (SEMA4C cytoplasmic domain-associated protein). It might also be responsible for axonal guidance, by forming clusters with SEMCAP-1, which in turn acts as ligand for receptors on adjacent cells. SEMA4C is highly expressed in adult brains, which suggests that this protein might be involved in synaptic plasticity. It might act as a bi-directional ligand for PSD-95 (postsynaptic density protein-95), in cortical neurons. Haploinsufficiency of this gene might be linked to Cri-du-chat syndrome, by causing abnormal brain development. SEMA4C might be involved in G-protein signaling through its interaction with SEMCAP-1. It is up-regulated in esophageal, gastric and rectal cancer, and might be responsible for the invasion and lymphatic metastasis of the same. It is involved in epithelial-mesenchymal transition in renal tubular epithelial cells, through TGF-β1-induced mitogen-activated protein kinase (MAPK) activation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72185

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuang-mei Ye et al.
Zhonghua yi xue za zhi, 92(28), 1954-1958 (2012-09-05)
To explore the expression and clinical significance of signal protein Sema4C in esophageal cancer, gastric cancer and rectal cancer. Fifty esophageal cancer, 75 gastric cancer, 50 rectal cancer and 20 corresponding normal mucous membrane specimens, collected during the period of
S Inagaki et al.
The Journal of biological chemistry, 276(12), 9174-9181 (2001-01-13)
Semaphorins are known to act as chemorepulsive molecules that guide axons during neural development. Sema4C, a group 4 semaphorin, is a transmembrane semaphorin of unknown function. The cytoplasmic domain of Sema4C contains a proline-rich region that may interact with some
Rui Zeng et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 26(4), 1149-1156 (2010-10-21)
The p38 mitogen-activated protein kinase (p38 MAPK) is an important intracellular signal transduction pathway involved in TGF-β1-induced epithelial-mesenchymal transition (EMT). Sema4C, a member of the semaphorin family, was found to be essential for the activation of p38 MAPK. However, the
A D Simmons et al.
Biochemical and biophysical research communications, 242(3), 685-691 (1998-02-17)
Cri-du-chat is a human contiguous gene deletion syndrome resulting from hemizygous deletions of chromosome 5p. Here we describe the isolation from within this interval of the human Semaphorin F (SEMAF) gene, a member of a family of proteins that has
L H Wang et al.
The Journal of biological chemistry, 274(20), 14137-14146 (1999-05-13)
M-SemF is a membrane-associated, neurally enriched member of the semaphorin family of axon guidance signals. We considered whether the cytoplasmic domain of M-SemF might possess a signaling function and/or might control the distribution of M-SemF on the cell surface. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service