Skip to Content
Merck
All Photos(2)

Key Documents

HPA011014

Sigma-Aldrich

Anti-GLIPR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GliPR 1, Anti-Glioma pathogenesis-related protein 1 precursor, Anti-RTVP-1 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

NEDFIKDCVRIHNKFRSEVKPTASDMLYMTWDPALAQIAKAWASNCQFSHNTRLKPPHKLHPNFTSLGENIWTGSVPIFSVSSAITNWYDEIQDYDF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLIPR1(11010)

General description

Glioma pathogenesis-related protein 1 (GLIPR1) is a multifunctional protein which was first recognized in human glioblastomas. It resides in endoplasmic reticulum (ER) and vesicles in cytoplasm. It is a transmembrane protein with its C-terminal spanning the membrane, and has a signal peptide at its N-terminal. It belongs to the CAP (cysteine-rich secretory proteins, antigen 5 and pathogenesis-related 1 proteins) family of proteins, and thus, contains the cysteine-rich CAP domain. GLIPR1 exists as three isoforms, namely, GLIPR1, GLIPR1-like 1 (GLIPR1L1) and GLIPR1-like 2 (GLIPR1L2). GLIPR1 and GLIPR1L2 have a wide range of tissue expression, whereas GLIPR1L1 is predominantly expressed in testis.

Immunogen

Glioma pathogenesis-related protein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Glioma pathogenesis-related protein 1 (GLIPR1) is involved in cell cycle, tumorigenesis and apoptosis. It acts as an oncogene in glioblastoma and gliomas, and is overexpressed in the same. It is also unregulated in acute myeloid leukemia (AML), and may facilitate the proliferation of AML cells. Its expression levels are positively correlated with metastasis of tumors in case of astrocytic brain malignancies. Studies suggest that GLIPR1 acts as a tumor suppressor gene in prostate cancer, and its expression is suppressed in prostate cancer cells as compared to normal prostate cells. The expression of this gene is induced by the tumor suppressor gene p53 as well as DNA damage-causing substances such as irradiation. It is an HIV-1 dependency factor (HDF), and it is overexpressed during initial stages of HIV-1 (human immunodeficiency virus-1) infection.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71999

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gianni Capalbo et al.
Oncology reports, 30(5), 2254-2262 (2013-09-07)
Glioma pathogenesis‑related protein 1 (GliPR1) is a pleiotropic protein involved in cell proliferation, tumor growth and apoptosis. The aim of the present study was to further characterize GliPR1 in regard to its subcellular localization and its overall effect on cellular
Yan-Hua Xiao et al.
Journal of cancer research and clinical oncology, 137(12), 1831-1840 (2011-09-17)
To identify methylation-silenced genes in acute myeloid leukemia (AML). Microarray analyses were performed in AML cell line HL-60 cells exposed to the demethylating agent 5-aza-2dC. The methylation status and expression of glioma pathogenesis-related protein 1 (GLIPR1), one of highly induced
Gianni Capalbo et al.
Retrovirology, 7, 26-26 (2010-04-02)
Previously, we showed that glioma pathogenesis related protein (GliPR) is induced in CEM T cells upon HIV-1 infection in vitro. To examine whether GliPR plays a role as HIV dependency factor (HDF), we tested the effect of GliPR suppression by
Oluwatoyin A Asojo et al.
Acta crystallographica. Section D, Biological crystallography, 67(Pt 10), 847-855 (2011-09-21)
Human glioma pathogenesis-related protein 1 (GLIPR1) is a membrane protein that is highly upregulated in brain cancers but is barely detectable in normal brain tissue. GLIPR1 is composed of a signal peptide that directs its secretion, a conserved cysteine-rich CAP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service