Skip to Content
Merck
All Photos(6)

Key Documents

HPA010734

Sigma-Aldrich

Anti-CD1A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD1a antigen antibody produced in rabbit, Anti-T-cell surface antigen T6/Leu-6 antibody produced in rabbit, Anti-T-cell surface glycoprotein CD1a precursor antibody produced in rabbit, Anti-hTa1 thymocyte antigen antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QNLVSGWLSDLQTHTWDSNSSTIVFLCPWSRGNFSNEEWKELETLFRIRTIRSFEGIRRYAHELQFEYPFEIQVTGGCELHSGKVSGSFLQLAYQGSDFVSFQNNSWLPYPVAGNMAKHFCKVLNQNQHEND

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD1A(909)

General description

CD1A (cluster of differentiation 1A) belongs to a family of antigen-presenting proteins called CD1. These are evolutionary conserved proteins which present lipids instead of peptide. Humans contain five CD1 genes namely, CD1A, CD1B, CD1C, CD1D and CD1E. CD1A is a transmembrane glycoprotein which is expressed on dendritic cells. It is the only isoform, in this family, to be expressed by Langerhans cells.

Immunogen

T-cell surface glycoprotein CD1a precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CD1 (cluster of differentiation 1) family of proteins is responsible for the capture and presentation of self and microbial glycolipids to the specific T cells. CD1A plays an important role in the host defense mechanism against microbial infection. It also facilitates early stages of immune response through CD1A-reactive T cells. This is achieved by inducing cytokine production and dendritic cell maturation, and activating cells such as NK (natural killer) cells and macrophages. The SNP rs411089, present in an intron in the CD1A gene is associated with increased susceptibility to tuberculosis, in Vietnamese population. Hence, it can be hypothesized that CD1A plays an important role in host defense against tuberculosis. The expression of this protein is also increased in monocyte culture derived from untreated multiple sclerosis (MS) patients, as opposed to expression in monocyte culture obtained from treated MS patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72090

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Salamero et al.
The Journal of investigative dermatology, 116(3), 401-408 (2001-03-07)
In this work, we studied the localization and traffic of CD1a molecules in human epidermal Langerhans cells and the ability of these cells to stimulate CD1a-restricted T cell clones. We found that CD1a was spontaneously internalized into freshly isolated Langerhans
C Seshadri et al.
Genes and immunity, 15(3), 195-198 (2014-02-07)
CD1 proteins are antigen-presenting molecules that evolved to present lipids rather than peptides to T cells. However, unlike major histocompatibility complex genes, CD1 genes show low rates of polymorphism and have not been clearly associated with human disease. We report
Michael S Vincent et al.
Journal of immunology (Baltimore, Md. : 1950), 175(10), 6344-6351 (2005-11-08)
Individual CD1-restricted T cells can recognize either endogenous or foreign lipid Ags, but the extent to which the same CD1-restricted TCR can react to both self and microbial lipids is unknown. In this study, we have identified CD1a-, CD1b-, and
S Bine et al.
Clinical and experimental immunology, 169(1), 10-16 (2012-06-08)
Studies of multiple sclerosis (MS) have concentrated mainly on antigen presentation of peptides derived from the myelin sheath, while the implication of lipid antigen has been less explored in this pathology. As the extracellular environment regulates expression of the lipid
Antonino Uncini et al.
Journal of the peripheral nervous system : JPNS, 16 Suppl 1, 48-51 (2011-07-01)
In the quest for susceptibility factors of inflammatory neuropathies, many genes implicated in the pathogenesis of autoimmune diseases have been investigated with negative or conflicting results. We studied, with a gene candidate approach, the CD1 system specialized in capturing and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service