Skip to Content
Merck
All Photos(2)

Key Documents

HPA010689

Sigma-Aldrich

Anti-PTGER3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PGE receptor, EP3 subtype, Anti-PGE2-R, Anti-Prostaglandin E2 receptor EP3 subtype, Anti-Prostanoid EP3 receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

MKETRGYGGDAPFCTRLNHSYTGMWAPERSAEARGNLTRPPGSGEDCGSVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTGER3(5733)

General description

PTGER3 (prostaglandin E receptor 3) gene encodes a seven-transmembrane-spanning protein with multiple C-terminal tails. The gene is mapped to human chromosome 1p31.2 and consists of 10 exons interspaced by nine introns. The TM domain and the 10 amino acid residues of the cytoplasmic tail are encoded by exon1 and 5′ 180-bp portion of exon 2.

Immunogen

Prostaglandin E2 receptor EP3 subtype recombinant protein epitope signature tag (PrEST)

Application

Anti-PTGER3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

PTGER3 (prostaglandin E receptor 3) gene encodes a G-protein coupled receptor that functions as a receptor for prostaglandin E2 (PGE2). It induces smooth-muscle contractility. It is moderately expressed in smooth muscle cells of the proximal, mid and distal ureter. It functions via cAMP response elements (CRE) and serves as a target for anti-inflammatory therapy in cutaneous inflammation. This receptor is mainly localized in primary sensory neurons and the spinal cord and functions in endogenous pain control. It has been found to be expressed in the joint nerves of patients with painful osteoarthritis. It plays a role in mediating antinociception during inflammation. It mediates contraction of human intercostal arteries by prostaglandin E2.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72461

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Longrois et al.
European journal of pharmacology, 681(1-3), 55-59 (2012-02-22)
Arterial vascularization of the spinal cord may be mechanically or functionally altered during thoraco-abdominal surgery/intravascular procedures. Increased arterial pressure has been shown to restore spinal perfusion and function probably by increasing the blood flow through the intercostal arteries. The regulation
Gabriel Natura et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(33), 13648-13653 (2013-08-02)
The pain mediator prostaglandin E2 (PGE2) sensitizes nociceptive pathways through EP2 and EP4 receptors, which are coupled to Gs proteins and increase cAMP. However, PGE2 also activates EP3 receptors, and the major signaling pathway of the EP3 receptor splice variants
M Kotani et al.
Genomics, 40(3), 425-434 (1997-03-15)
Prostaglandin EP3 receptor subtype is a seven-membrane-spanning protein with multiple C-terminal tails generated by alternative mRNA splicing. We report here the structural organization of the human EP3 gene (PTGER3). The human EP3 gene spanned more than 80 kb and was
Matthias Oll et al.
BMC urology, 12, 35-35 (2012-12-12)
Prostaglandins play an important role in ureteral obstruction, but the detailed expression profiles of the prostaglandin receptors (PTGER1, PTGER2, PTGER3, PTGER4, PTGFR) remain unknown in the different parts of the human ureter. The expression pattern of PTGER1, PTGER2, PTGER3, PTGER4

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service