Skip to Content
Merck
All Photos(4)

Key Documents

HPA010574

Sigma-Aldrich

Anti-SYT4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HsT1192, Anti-KIAA1342, Anti-synaptotagmin IV

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SNKTPPYKFVHVLKGVDIYPENLNSKKKFGADDKNEVKNKPAVPKNSLHLDLEKRDLNGNFPKTNLKPGSPSDLENATPKLFLEGEKESVSPESLKSSTSLTSEEKQEKLGTLFFSLEYNFERKAFVVNIKEARGLPAMDEQSMT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYT4(6860)

General description

SYT4 (synaptotagmin 4) gene is localized on human chromosome 18q12.3. Its mRNA is predominantly expressed in hippocampus of human brain, and shows lesser expression in amygdala and thalamus. This protein is a member of the SYT protein family. In PC12 (pheochromocytoma) cell line, it is expressed in endoplasmic reticulum-Golgi complexes and secretory granules. In culture neurons, this protein localizes to axonal processes. This protein forms a part of neuroendocrine system.

Immunogen

synaptotagmin IV recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SYT4 (synaptotagmin 4) protein is thought to play a role in human brain functioning and neurological disorders. SYT4 null mice die shortly after birth. It is an atypical regulator of synaptic exocytosis, and also negatively controls the exocytosis of oxytocin. In dietary obesity, there is an increased vesicle binding of SYT4, which leads to increased negative regulation of oxytocin exocytosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71943

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo Zhang et al.
Neuron, 69(3), 523-535 (2011-02-15)
Hypothalamic neuropeptides play essential roles in regulating energy and body weight balance. Energy imbalance and obesity have been linked to hypothalamic signaling defects in regulating neuropeptide genes; however, it is unknown whether dysregulation of neuropeptide exocytosis could be critically involved.
G D Ferguson et al.
The Journal of biological chemistry, 275(47), 36920-36926 (2000-08-12)
Rat synaptotagmin IV (SYT IV) is a depolarization-inducible synaptic vesicle protein. SYT IV homozygous mutant mice are viable and have deficits in fine motor coordination and some forms of memory. In this study, we report the identification of a human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service