Skip to Content
Merck
All Photos(2)

Key Documents

HPA010510

Sigma-Aldrich

Anti-CNMD antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BRICD3, Anti-CHM-I, Anti-CHM1, Anti-LECT1, Anti-MYETS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

NGITGIRFAGGEKCYIKAQVKARIPEVGAVTKQSISSKLEGKIMPVKYEENSLIWVAVDQPVKDNSFLSSKVLELCGDLPIFWLKPTYPKEIQRERREVVRKIVPTTTKRPHSGPRSNPGAGRLNNETRPSVQ

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LECT1(11061)

General description

LECT1 (leukocyte cell derived chemotaxin 1) commonly known as chondromodulin-I (CHM-1), is a glycoprotein which is specific to cartilage. This gene is localized to human chromosome 13q14-21.

Immunogen

chondromodulin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LECT1 (leukocyte cell derived chemotaxin 1) is involved in the development of precursor of tumor cells and is associated with site of tumor development in chondrosarcomas. This protein is linked with chondrocyte growth and prevents the formation of tube in endothelial cells. It inhibits angiogenesis in cartilage, and it inhibits tumor growth in HT-29 colon adenocarcinoma cell line. Thus, it might have a therapeutic potential in case of solid tumors. Along with endostatin, it is responsible for maintaining the vascularity of cartilage. It preserves the avascularity of the inner meniscus by functioning as an anti-angiogenic factor.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71960

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoki Aoyama et al.
Cancer letters, 204(1), 61-68 (2004-01-28)
We investigated the expression of the Chondromodulin-I (ChM-I) gene, a putative tumor suppressor gene in cartilaginous tumors, by quantitative RT-PCR in 15 chondrosarcomas (CSs). Eight CSs expressed the ChM-I gene at the level higher than those in articular cartilage (positive
I Yanagihara et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 15(3), 421-429 (2000-04-06)
Chondromodulin-1 (ChM-1) is a cartilage-specific glycoprotein that stimulates the growth of chondrocytes and inhibits the tube formation of endothelial cells. To clarify the tissue-specific expression and the role of ChM-1 in pathophysiological conditions, we analyzed the structure of the human
Masataka Fujii et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 31(4), 538-543 (2012-11-13)
The meniscus is a fibrocartilaginous tissue that plays an important role in controlling complex biomechanics of the knee. A perimeniscal capillary plexus supplies the outer meniscus, whereas the inner meniscus is composed of avascular tissue. Anti-angiogenic molecules, such as chondromodulin-I
T Hayami et al.
FEBS letters, 458(3), 436-440 (1999-11-26)
Chondromodulin-I (ChM-I) was previously identified as an angiogenesis inhibitor in cartilage. Here, we demonstrated that the level of ChM-I transcripts was substantially reduced to 100 or even less in the lower-grade chondrosarcomas, in articular cartilage or other benign cartilage tumors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service