Skip to Content
Merck
All Photos(6)

Key Documents

HPA009173

Sigma-Aldrich

Anti-CDHR5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-μ-Protocadherin, Anti-MUCDHL, Anti-MUPCDH, Anti-Mucin and cadherin-like protein precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

CSVNKDIFEVEENTNVTEPLVDIHVPEGQEVTLGALSTPFAFRIQGNQLFLNVTPDYEEKSLLEAQLLCQSGGTLVTQLRVFVSVLDVNDNAPEFPFKTKEIRVEEDTKVNSTVIPETQLQAEDRDKDDILFYTLQEMTAGASDYFSLVS

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUPCDH(53841)

Looking for similar products? Visit Product Comparison Guide

General description

CDHR5 (cadherin related family member 5) is a protocadherin (PCDH) family member, and is also known as PCDH24. It might be a constituent of enterocyte microvillus. It is a poorly characterized protein. It is a type I transmembrane protein, which contains the characteristic domains of the cadherin protein family- extracellular EC repeats. This gene localizes to human chromosome 11p15.5.

Immunogen

Mucin and cadherin-like protein precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CDHR5 (cadherin related family member 5) is localized to the brush border, and might be involved in the adhesion of microvilli, eventually resulting in the tight packing of fully differentiated enterocytes. It interacts in a trans-fashion with MLPCDH (mucin-like protocadherin) to form Ca2+-dependent adhesion complexes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71709

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felix Marius Bläsius et al.
Oncotarget, 8(43), 75076-75086 (2017-11-02)
Reduced expression of Cadherin-Related Family Member 5 (CDHR5) was recently found implied in carcinogenesis of colon cancer, but its role in other tumors is unknown. We aimed to analyze the expression of CDHR5 in different subtypes of renal cell carcinoma.
Scott W Crawley et al.
Cell, 157(2), 433-446 (2014-04-15)
Transporting epithelial cells build apical microvilli to increase membrane surface area and enhance absorptive capacity. The intestinal brush border provides an elaborate example with tightly packed microvilli that function in nutrient absorption and host defense. Although the brush border is
Michael Goldberg et al.
American journal of physiology. Renal physiology, 283(3), F454-F463 (2002-08-09)
We recently cloned mu-protocadherin, a developmentally regulated cell adhesion molecule that contains an extracellular region with four cadherin-like ectodomains and a triply repeating mucin domain in its longer isoform. Expression of mu-protocadherin in L929 cells resulted in cellular aggregation, confirming
Bernadette Mödl et al.
EMBO reports, 24(10), e57084-e57084 (2023-09-11)
Intestinal epithelial cells are covered by the brush border, which consists of densely packed microvilli. The Intermicrovillar Adhesion Complex (IMAC) links the microvilli and is required for proper brush border organization. Whether microvillus crosslinking is involved in the intestinal barrier
Russell E McConnell et al.
American journal of physiology. Gastrointestinal and liver physiology, 300(5), G914-G926 (2011-02-19)
The brush border domain at the apex of intestinal epithelial cells is the primary site of nutrient absorption in the intestinal tract and the primary surface of interaction with microbes that reside in the lumen. Because the brush border is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service