Skip to Content
Merck
All Photos(6)

Documents

HPA009087

Sigma-Aldrich

Anti-CLCC1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Mid-1-related chloride channel 1 isoform 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PPQALRPRDRRRQEEIDYRPDGGAGDADFHYRGQMGPTEQGPYAKTYEGRREILRERDVDLRFQTGNKSPEVLRAFDVPDAEAREHPTVVPSHKSPVLDTKPKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLCC1(23155)

Looking for similar products? Visit Product Comparison Guide

General description

CLCC1 (chloride channel CLIC like 1) is an intracellular chloride channel, which belongs to the CLIC channel family. This protein is composed of 551 amino acids, and is thought to have four transmembrane domains. Its mRNA is expressed at high levels in testis, and is moderately expressed in brain, heart, lung, kidney, liver and spleen. It localizes to Golgi bodies, nucleus and ER (endoplasmic reticulum), and is related to Mid-1 protein of Saccharomyces cerevisiae. This gene localizes to human chromosome 1.

Immunogen

Mid-1-related chloride channel 1 isoform 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CLCC1 (chloride channel CLIC like 1) functions as an anion channel. Inactivation of this protein in ER (endoplasmic reticulum) results in increase of HSP70 (heat shock protein) family member GRP78. This results in the accumulation of ubiquitinated proteins in neurons leading to degeneration. Thus, chloride homeostasis is essential for the proper protein-folding functionality of ER.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71867

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yichang Jia et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(7), 3001-3009 (2015-02-24)
Folding of transmembrane and secretory proteins occurs in the lumen of the endoplasmic reticulum (ER) before transportation to the cell surface and is monitored by the unfolded protein response (UPR) signaling pathway. The accumulation of unfolded proteins in the ER
M Nagasawa et al.
The Journal of biological chemistry, 276(23), 20413-20418 (2001-03-30)
MID-1 is a Saccharomyces cerevisiae gene encoding a stretch-activated channel. Using MID-1 as a molecular probe, we isolated rat cDNA encoding a protein with four putative transmembrane domains. This gene encoded a protein of 541 amino acids. We also cloned
Marguerite Ryan Irvin et al.
Pharmacogenetics and genomics, 21(12), 798-807 (2011-09-08)
Genetic studies may help explain abnormalities of fat distribution in HIV-infected patients treated with antiretroviral therapy (ARV). Subcutaneous adipose tissue (SAT) volume measured by MRI in the leg, the lower trunk, the upper trunk, and the arm was examined in
Xinhua Li et al.
Annual review of physiology, 64, 609-633 (2002-02-05)
Hepatocytes possess chloride channels at the plasma membrane and in multiple intracellular compartments. These channels are required for cell volume regulation and acidification of intracellular organelles. Evidence also supports a role of chloride channels in modulation of apoptosis and cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service