Skip to Content
Merck
All Photos(4)

Key Documents

HPA009073

Sigma-Aldrich

Anti-AATK antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AATYK, Anti-AATYK1, Anti-KIAA0641, Anti-LMR1, Anti-LMTK1, Anti-PPP1R77

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

TSGIFTDTSSDGLQARRPDVVPAFRSLQKQVGTPDSLDSLDIPSSASDGGYEVFSPSATGPSGGQPRALDSGYDTENYESPEFVLKEAQEGCEPQAFAELASEGEGPGPETRLSTSLSGLNEKNPYRDSA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AATK(9625)

General description

AATK (apoptosis-associated tyrosine kinase) was recently identified in 32Dcl3 mouse myeloid cells, which were subjected to IL (interleukin)-3 withdrawal and undergoing apoptosis. This protein is composed of a putative tyrosine kinase domain, and has multiple Src homology 2 (SH2) and (SH3)-binding motifs. This protein is predominantly expressed in brain, and is localized to the cytoplasm. It is an active, non-receptor kinase. This protein is composed of 1316 amino acids and has a molecular weight of 145kDa. The N-terminal of this protein shows high homology to tyrosine kinases. This gene is localized to human chromosome 17q25.3.

Immunogen

Serine/threonine-protein kinase LMTK1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AATK (apoptosis-associated tyrosine kinase) faciliates the differentiation of neurons when stimulated by all-trans retinoic acid (RA), 12-O-Tetradecanoyl phorbol 13-acetate (TPA) and IGF-I (insulin like growth factor-I). it induces neuronal differentiation even in the absence of these agents. It is an essential element for the induction of apoptosis and growth arrest in myeloid precursor cells. The expression of AATK mRNA was significantly elevated in cultured cerebral granule cells during apoptosis, suggesting this protein plays an important role in neuronal apoptosis. In developing neurons, this protein is involved in the extension of neurites through its tyrosine kinase activity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71460

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Gaozza et al.
Oncogene, 15(25), 3127-3135 (1998-01-28)
Apoptosis, or programmed cell death, is a process where developmental or environmental stimuli activate a genetic program to implement a series of events that culminate in cell death. To study the nature of genes that are induced during the apoptotic
M Raghunath et al.
Brain research. Molecular brain research, 77(2), 151-162 (2000-06-06)
Apoptosis Associated Tyrosine Kinase (AATYK), a novel protein recently isolated from differentiating 32D mouse myeloid cells, contains a putative tyrosine kinase domain and several binding motifs for src homology 2 (SH-2) and src homology 3 (SH-3) domain containing proteins. We
N Seki et al.
Journal of human genetics, 44(2), 141-142 (1999-03-20)
Here, we report on the chromosomal location of the human apoptosis-associated tyrosine kinase gene. Based on polymerase chain reaction analysis with a human/rodent monochromosomal hybrid cell panel and fluorescence in situ hybridization, the gene was mapped on 25.3 region of
Mineko Tomomura et al.
Brain research. Molecular brain research, 112(1-2), 103-112 (2003-04-03)
Apoptosis-associated tyrosine kinase (AATYK) is a non-receptor type tyrosine kinase that is predominantly expressed in adult mouse brain. Although it is also expressed in developing brains, its expression pattern and physiological functions are unclear. In the present study, we analyzed
M Tomomura et al.
Oncogene, 20(9), 1022-1032 (2001-04-21)
We isolated three related cDNA clones from a mouse cerebellar library; the type I cDNA was identical to the gene encoding the apoptosis-associated tyrosine kinase (AATYK), whose expression in myeloid precursor cells is increased during growth arrest or apoptosis. Low

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service