Skip to Content
Merck
All Photos(4)

Key Documents

HPA009025

Sigma-Aldrich

Anti-FAM213A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti- MGC4248, Anti-C10orf58, Anti-PAMM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NTDVFLSKPQKAALEYLEDIDLKTLEKEPRTFKAKELWEKNGAVIMAVRRPGCFLCREEAADLSSLKSMLDQLGVPLYAVVKEHIRTEVKDFQPYFKGE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

FAM213A (family with sequence similarity 213 member A) is a perirdeoxin-like protein, which contains a CXXC motif. Upon stimulation by M-CSF (macrophage-colony stimulating factor) and RANKL (receptor activator of nuclear factor κ-B ligand), this protein is expressed by bone marrow monocytes, and hence, the name PAMM (peroxiredoxin (PRX)-like 2 activated in M-CSF stimulated monocytes).it is also expressed in brain, liver and kidney.

Immunogen

Uncharacterized protein C10orf58 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-C10orf58 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

FAM213A (family with sequence similarity 213 member A) plays a key role in osteoclast differentiation, where its expression is reduced by RANKL (receptor activator of nuclear factor κ-B ligand), which results in an elevation of ROS (reactive oxygen species). Thus, it is a redox regulatory protein, and might also be involved in bone resorption, thus, playing a part in maintaining bone mass.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71870

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Guo et al.
The Biochemical journal, 472(3), 309-318 (2015-10-07)
Macrophages within adipose tissue play a key role in mediating inflammatory responses in adipose tissue that are associated with obesity-related metabolic complications. In an effort to identify novel proteins secreted from adipocytes that may negatively regulate macrophage inflammation, we found
Yan Xu et al.
Antioxidants & redox signaling, 13(1), 27-37 (2009-12-03)
The central role of reactive oxygen species (ROS) in osteoclast differentiation and in bone homeostasis prompted us to characterize the redox regulatory system of osteoclasts. In this report, we describe the expression and functional characterization of PAMM, a CXXC motif-containing
Yan Xu et al.
Redox biology, 6, 446-453 (2015-09-25)
Reactive oxygen species (ROS) play a central role in estrogen deficiency-induced bone loss. We previously identified and characterized a novel member of the Peroxiredoxin (PRX) like 2 family that we called PAMM: Peroxiredoxin Activated in M-CSF stimulated Monocytes, a redox

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service