Skip to Content
Merck
All Photos(2)

Key Documents

HPA008904

Sigma-Aldrich

Anti-ADGRA3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ38547, Anti-GPR125, Anti-PGR21

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Check Cart for Availability


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

DIFRGLTNLVRLNLSGNLFSSLSQGTFDYLASLRSLEFQTEYLLCDCNILWMHRWVKEKNITVRDTRCVYPKSLQAQPVTGVKQELLTCDPPLELPSFYMTPSHRQVVFEGDSLPFQCMASYIDQDMQVLWYQDGRI

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR125(166647)

General description

GPR125 (G protein-coupled receptor 125) is a candidate marker for human spermatogonial stem cells (SSCs). It is expressed in adult mice testis and is found in long-term pre-pubertal testicular cell cultures that contain SSCs. It is an orphan GPCR. It is a homolog of TEM5 protein, and thus, is also known as TEM5-like protein. This protein is composed of 1205 amino acids, and shares homology to B family of GPCRs. This gene is localized to human chromosome 4p15.

Immunogen

adhesion G protein-coupled receptor A3

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The function of GPR125 (G protein-coupled receptor 125) is not very clear yet. In conjunction with other stem cell markers, this protein might aid in recognizing spermatogonial subpopulations in both young and old donors. Human discs large tumor suppressor gene (hDlg) scaffolds TEM5 and GPR125 proteins in endothelial cells during angiogenesis and neoangiogensis in cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71172

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Sachs et al.
Andrology, 2(4), 607-614 (2014-06-07)
The optimal markers for human spermatogonial stem cells (SSCs) are not known. Among the genes recently linked to SSCs in mice and other animals are the basic helix-loop-helix transcription factor ID4 and the orphan G-protein-coupled receptor GPR125. While ID4 and
A Christoforou et al.
Molecular psychiatry, 12(11), 1011-1025 (2007-04-26)
Several independent linkage studies have identified chromosome 4p15-p16 as a putative region of susceptibility for bipolar disorder (BP), schizophrenia (SCZ) and related phenotypes. Previously, we identified two subregions (B and D) of the 4p15-p16 region that are shared by three
Yasunori Yamamoto et al.
Oncogene, 23(22), 3889-3897 (2004-03-17)
The human homologue of the Drosophila discs large tumor suppressor gene (hDlg) is a member of the membrane-associated guanylate kinase family with three PSD-95/Dlg/ZO-1 (PDZ) domains. hDlg has been shown to bind tumor suppressor proteins, adenomatous polyposis coli (APC) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service