Skip to Content
Merck
All Photos(6)

Key Documents

HPA008819

Sigma-Aldrich

Anti-PEBP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HCNPpp, Anti-Neuropolypeptide h3, Anti-PEBP-1, Anti-Phosphatidylethanolamine-binding protein 1, Anti-Prostatic-binding protein, Anti-RKIP, Anti-Raf kinase inhibitor protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

PTQVKNRPTSISWDGLDSGKLYTLVLTDPDAPSRKDPKYREWHHFLVVNMKGNDISSGTVLSDYVGSGPPKGTGLHRYVWLVYEQDRPLKCDEPILSNRSGDHRGKFKVASFRKKYELRAPVAGTCYQAEWDDYVPKLY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PEBP1(5037)

General description

PEBP1 (phosphatidylethanolamine binding protein 1), also called Raf kinase inhibitory protein (RKIP), is a member of the PEBP class of proteins. This class was initially identified as the class of proteins specifically interacting with phosphatidylethanolamine phospholipid. This class contains two members- PEBP1 and PEBP4. This protein exists in multiple conformations and has a molecular weight of 21kDa.

Immunogen

Phosphatidylethanolamine-binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PEBP1 (phosphatidylethanolamine binding protein 1) binds with and prevents the phosphorylation of Raf-1 protein, which is a suppressor of mitogen activated protein kinase (MAPK) pathway. Thus, it indirectly prevents the activation of MAPK-extracellular signal-regulated kinase (ERK) signaling, and functions as an anti-inflammatory protein. Therefore, the interaction of PEBP1 protein with phospholipids, proteins and fatty acids might have a role in asthma and diabetes. It also acts as a negative regulator of β-adrenergic signaling and NFκB signaling, and acts as the regulator of the mammalian kinome. Inactivation of this gene might result in metastatic cancer, chromosomal abnormalities, systemic inflammatory response syndrome, cardiac and Alzheimer′s disease. Its loss of function also leads to colonization by Helicobacter pylori, which leads to the death of gastric epithelial cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71549

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John J Skinner et al.
Critical reviews in oncogenesis, 19(6), 483-488 (2015-01-20)
Raf kinase inhibitory protein (RKIP) is a highly conserved regulator of many signaling networks whose loss or inactivation can lead to a variety of disease states. The multifaceted roles played by RKIP are enabled by an allosteric structure that is
Jinming Zhao et al.
Critical reviews in oncogenesis, 19(6), 497-504 (2015-01-20)
The inflammatory response plays an important role in host defense and maintenance of homeostasis, while imbalances in these responses can also lead to pathologic disease processes. Emerging data show that RKIP interacts with multiple signaling molecules that may potentiate multiple
Sahl Khalid Bedri et al.
PloS one, 14(5), e0217208-e0217208 (2019-05-30)
Multiple sclerosis (MS) treatment options have improved significantly over the past decades, but the consequences of MS can still be devastating and the needs for monitoring treatment surveillance are considerable. In the current study we used affinity proteomics technology to
Pascal Schlage et al.
Molecular & cellular proteomics : MCP, 14(12), 3234-3246 (2015-10-18)
Matrix metalloproteinases (MMPs) are important players in skin homeostasis, wound repair, and in the pathogenesis of skin cancer. It is now well established that most of their functions are related to processing of bioactive proteins rather than components of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service