Skip to Content
Merck
All Photos(5)

Key Documents

HPA008758

Sigma-Aldrich

Anti-THAP12 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DAP4, Anti-P52rIPK, Anti-PRKRIR, Anti-THAP0

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

NPHSRHRKRIKELSEDEIRTLKQKKIDETSEQEQKHKETNNSNAQNPSEEEGEGQDEDILPLTLEEKENKEYLKSLFEILILMGKQNIPLDGHEADEIPEGLFTPDNFQALLECRINSGEEVLRKRFE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKRIR(5612)

General description

PRKRIR (protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor)) is a P58IPK-interacting protein, which has a wide range of tissue expression. It is a novel member of the PKR (protein kinase R)-regulatory cascade. This gene is localized to human chromosome 11q13.5.

Immunogen

THAP domain containing 12 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PRKRIR (protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor)) suppressed the function of P58IPK in an in vitro PKR (protein kinase R)-regulatory assay. This inhibition results in increased PKR activity, forming a connection among interferon (IFN), stress and growth-regulatory pathways. This protein regulates protein stability and acts as a positive regulator of RIG-I (retinoic acid-inducible gene 1) protein. The latter protein recognizes foreign or viral pathogens and activates type I interferon (IFN) production.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71535

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hesung Now et al.
Biochemical and biophysical research communications, 413(3), 487-493 (2011-09-14)
The cellular RIG-I-like receptor (RLR) senses pathogenic RNA molecular patterns and transmits signals for type I interferon (IFN) production. It acts as a center for antiviral responses, and large numbers of RIG-I (retinoic acid inducible gene-I) interacting proteins are identified
M Gale et al.
Molecular and cellular biology, 18(2), 859-871 (1998-02-03)
The cellular response to environmental signals is largely dependent upon the induction of responsive protein kinase signaling pathways. Within these pathways, distinct protein-protein interactions play a role in determining the specificity of the response through regulation of kinase function. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service