Skip to Content
Merck
All Photos(3)

Documents

HPA008445

Sigma-Aldrich

Anti-YWHAE antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-14-3-3 protein epsilon, Anti-14-3-3E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

MDDREDLVYQAKLAEQAERYDEMVESMKKVAGMDVELTVEERNLLSVAYKNVIGARRASWRIISSIEQKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YWHAE(7531)

General description

Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein-ε (YWHAE) is one of the adaptor proteins that interacts with phosphorylated serine or threonine residue of target proteins. The gene encoding this protein is present on chromosome 17p13.

Immunogen

14-3-3 protein epsilon recombinant protein epitope signature tag (PrEST)

Application

Anti-YWHAE antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-YWHAE antibody produced in rabbit can also be used for immunoprecipitation.

Biochem/physiol Actions

Tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein-ε (YWHAE) binds to cyclin-dependent kinase 5 (CDK5)-phosphorylated nudE neurodevelopment protein 1-like 1 (NUDEL) and maintains its phosphorylation state. NUDEL is a cytoplasmic regulator of neuronal migration. YWHAE is required for neuronal development as it aids in the transport of the NUDEL complex. It has protective and preventive effect on schizophrenia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71114

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masashi Ikeda et al.
Human molecular genetics, 17(20), 3212-3222 (2008-07-29)
Schizophrenia is a complex mental disorder with a fairly high degree of heritability. Although the causes of schizophrenia remain unclear, it is now widely accepted that it is a neurodevelopmental and neurodegenerative disorder involving disconnectivity and disorder of the synapses.
Wen-Bin Ou et al.
Oncogenesis, 10(5), 37-37 (2021-05-06)
Endometrial stromal sarcoma (ESS) is the second most common subtype of uterine mesenchymal cancer, after leiomyosarcoma, and oncogenic fusion proteins are found in many ESS. Our previous studies demonstrated transforming properties and diagnostic relevance of the fusion oncoprotein YWHAE-NUTM2 in
Assignment of the human 14-3-3 epsilon isoform (YWHAE) to human chromosome 17p13 by in situ hybridization.
S C Luk et al.
Cytogenetics and cell genetics, 78(2), 105-106 (1997-01-01)
Cheng-Han Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(3), 929-934 (2012-01-10)
14-3-3 proteins are ubiquitously expressed regulators of various cellular functions, including proliferation, metabolism, and differentiation, and altered 14-3-3 expression is associated with development and progression of cancer. We report a transforming 14-3-3 oncoprotein, which we identified through conventional cytogenetics and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service