Skip to Content
Merck
All Photos(1)

Documents

HPA008312

Sigma-Aldrich

Anti-PML antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Probable transcription factor PML antibody produced in rabbit, Anti-RING finger protein 71 antibody produced in rabbit, Anti-Tripartite motif-containing protein 19 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

ISGFLAALPLIRERVPGASSFKLKNLAQTYLARNMSERSAMAAVLAMRDLCRLLEVSPGPQLAQHVYPFSSLQCFASLQPLVQAAVLPRAEARLLALHNVSFMELLSAHRRDRQGGLKKYSRYLSLQTTTLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PML(5371)

General description

Promyelocytic leukemia (PML) belongs to the B-box family of proteins.

Immunogen

Probable transcription factor PML recombinant protein epitope signature tag (PrEST)

Application

Anti-PML antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Promyelocytic leukemia (PML) plays an important role as a cell growth and tumor suppressor protein. It is essential for multiple apoptotic signals. PML is present in speckled nuclear structures called PML nuclear bodies (NB). In mature activated splenic lymphocytes, a protein called Daxx (death domain-associated protein) is upregulated and it accumulates in the PML-NBs. Here, PML and Daxx physically interact and take part in a pathway for apoptosis. It also acts as a ligand-dependent coactivator of retinoid X receptor α (RXR-α)/retinoic acid receptor α (RAR-α).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70832

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Zhong et al.
Nature genetics, 23(3), 287-295 (1999-12-28)
PML and Tif1a are fused to RARA and Braf, respectively, resulting in the production of PML-RARalpha and Tif1alpha-B-Raf (T18) oncoproteins. Here we show that PML, Tif1alpha and RXRalpha/RARalpha function together in a transcription complex that is dependent on retinoic acid
Wesley H Stepp et al.
mBio, 4(6), e00845-e00813 (2013-11-07)
Most DNA viruses associate with, and reorganize, nuclear domain 10 (ND10) bodies upon entry into the host nucleus. In this study, we examine the roles of the ND10 components PML, Sp100, and Daxx in the establishment of human papillomavirus type
T Cao et al.
Journal of cell science, 111 ( Pt 10), 1319-1329 (1998-05-07)
The ret finger protein (rfp) is a member of the B-box zinc finger gene family many of which may function in growth regulation and in the appropriate context become oncogenic. Members of this family are nuclear proteins that possess a
S Zhong et al.
The Journal of experimental medicine, 191(4), 631-640 (2000-02-24)
The promyelocytic leukemia protein (PML) gene of acute promyelocytic leukemia (APL) encodes a cell growth and tumor suppressor essential for multiple apoptotic signals. Daxx was identified as a molecule important for the cytoplasmic transduction of the Fas proapoptotic stimulus. Here
M Campagna et al.
Cell death and differentiation, 18(1), 72-79 (2010-06-26)
SIRT1, the closest mammalian homolog of yeast Sir2, is an NAD(+)-dependent deacetylase with relevant functions in cancer, aging, and metabolism among other processes. SIRT1 has a diffuse nuclear localization but is recruited to the PML nuclear bodies (PML-NBs) after PML

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service