Skip to Content
Merck
All Photos(5)

Key Documents

HPA008238

Sigma-Aldrich

Anti-MCPH1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Microcephalin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AVGLKSTQNKGTTSKISNSSEGEAQSEHEPCFIVDCNMETSTEEKENLPGGYSGSVKNRPTRHDVLDDSCDGFKDLIKPHEELKKSGRGKKPTRTLVMTSMPSEKQNVVIQVVDKLKGFSIAPDVCETTTHVLSGKPLRTLNVLLGI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCPH1(79648)

General description

The gene encoding microcephalin 1 (MCPH1) is present on chromosome 8p23. It contains twin carboxyl-terminal BRCA1 C-terminus (BRCT) domains.

Immunogen

Microcephalin recombinant protein epitope signature tag (PrEST)

Application

Anti-MCPH1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Microcephalin 1 (MCPH1) is involved in DNA damage-induced cellular responses by regulating the pathways connected with breast cancer 1 (BRCA1) and Chk1 (a serine/threonine-protein kinase). It has also been shown that it regulates brain size during development and is involved in the evolutionary enlargement of the brain from primates to humans. MCPH1 also interacts with Chk1 and has an important role downstream of Chk1, in the ataxia telangiectasia and rad3-related protein (ATR)-signaling pathway. It maintains inhibitory cyclin-dependent kinase 1 (Cdk1) phosphorylation, hence preventing its premature entry into mitosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71533

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick D Evans et al.
Human molecular genetics, 13(11), 1139-1145 (2004-04-02)
The defining process in the evolution of primates and particularly humans is the dramatic expansion of the brain. While many types of genes could potentially contribute to this process, genes that specifically regulate brain size during development may be especially
Gemma K Alderton et al.
Nature cell biology, 8(7), 725-733 (2006-06-20)
Ataxia-telangiectasia mutated and Rad3 related (ATR)-Seckel syndrome and autosomal recessive primary microcephaly (MCPH) syndrome share clinical features. RNA interference (RNAi) of MCPH1 have implicated the protein it encodes as a DNA-damage response protein that regulates the transcription of Chk1 and
Xingzhi Xu et al.
The Journal of biological chemistry, 279(33), 34091-34094 (2004-06-29)
Microcephalin (MCPH1) is the first gene identified among at least six loci that contribute to the autosomal recessive disease, primary microcephaly. MCPH1, like NFBD1/MDC1, 53BP1, and BRCA1, encodes a protein with twin carboxyl-terminal BRCT domains (PTCB). Here, we report that
Thejaswini Venkatesh et al.
PloS one, 8(3), e54643-e54643 (2013-03-09)
Mutations in the MCPH1 (microcephalin 1) gene, located at chromosome 8p23.1, result in two autosomal recessive disorders: primary microcephaly and premature chromosome condensation syndrome. MCPH1 has also been shown to be downregulated in breast, prostate and ovarian cancers, and mutated
Andrew P Jackson et al.
American journal of human genetics, 71(1), 136-142 (2002-06-05)
Primary microcephaly (MIM 251200) is an autosomal recessive neurodevelopmental condition in which there is a global reduction in cerebral cortex volume, to a size comparable with that of early hominids. We previously mapped the MCPH1 locus, for primary microcephaly, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service