Skip to Content
Merck
All Photos(4)

Key Documents

HPA007728

Sigma-Aldrich

Anti-HGS antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Hepatocyte growth factor-regulated tyrosine kinase substrate, Anti-Hrs, Anti-Protein pp110

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PHVALYALEVMESVVKNCGQTVHDEVANKQTMEELKDLLKRQVEVNVRNKILYLIQAWAHAFRNEPKYKVVQDTYQIMKVEGHVFPEFKESDAMFAAERAPDWVDAEECHRCRVQFGVMTRKHHCRACGQIFCGKCS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HGS(9146)

General description

HGS (hepatocyte growth factor-regulated tyrosine kinase substrate) protein has a molar mass of 110kDa and contains a double zinc-finger motif. It is localized in the cytoplasm. It is a 777-amino-acid protein with 93% homology to mouse Hrs. The gene is mapped to human chromosome 17q25.

Immunogen

Hepatocyte growth factor-regulated tyrosine kinase substrate recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HGS (hepatocyte growth factor-regulated tyrosine kinase substrate) gene encodes a FYVE domain protein localized to early endosomes. It functions in membrane trafficking of various molecules. It recruits Smad2 and Smad3 to the activin receptor complex in cooperation with SARA and regulates Smad signaling involved in mouse early embryonic development. It functions in the regulation of endosomal sorting and targets proteins for lysosome-dependent degradation. Rapid phosphorylation of Hgs is stimulated by IL-2, granulocyte-macrophage colony-stimulating factor and hepatocyte growth factor. It is associated with STAM, a signal-transducing adaptor molecule, and suppresses its function in cell growth signaling mediated by the cytokines.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86878

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Miura et al.
Molecular and cellular biology, 20(24), 9346-9355 (2000-11-30)
Smad proteins are effector molecules that transmit signals from the receptors for the transforming growth factor beta (TGF-beta) superfamily to the nucleus; of the Smad proteins, Smad2 and Smad4 are essential components for mouse early embryogenesis. We demonstrated that Hgs
H Asao et al.
The Journal of biological chemistry, 272(52), 32785-32791 (1998-01-31)
We previously reported a new type of signal-transducing adaptor molecule, STAM, which was shown to be involved in cytokine-mediated intracellular signal transduction. In this study, we molecularly cloned a 110-kDa phosphotyrosine protein inducible by stimulation with interleukin 2 (IL-2). The
L Lu et al.
Gene, 213(1-2), 125-132 (1998-06-19)
Hrs is a 115kDa zinc finger protein which is rapidly tyrosine phosphorylated in cells stimulated with various growth factors. We previously purified the protein from a mouse cell line and cloned its cDNA. In the present study, we cloned a
Benoît T Roux et al.
Scientific reports, 7(1), 12389-12389 (2017-09-30)
Calcitonin receptor-like receptor (CLR) and the receptor activity-modifying protein 2 (RAMP2) comprise a receptor for adrenomedullin (AM). Although it is known that AM induces internalization of CLR•RAMP2, little is known about the molecular mechanisms that regulate the trafficking of CLR•RAMP2.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service