Skip to Content
Merck
All Photos(9)

Key Documents

HPA006759

Sigma-Aldrich

Anti-GATAD2A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GATA zinc finger domain-containing protein 2A antibody produced in rabbit, Anti-Hp66α antibody produced in rabbit, Anti-Transcriptional repressor p66 α antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RRKLAFRSGEARDWSNGAVLQASSQLSRGSATTPRGVLHTFSPSPKLQNSASATALVSRTGRHSERTVSAGKGSATSNWKKTPLSTGGTLAFVSPSLAVHKSSSAVDRQREYLLDMIPPRSIP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GATAD2A(54815)

General description

GATA zinc finger domain containing 2A (GATAD2A) comprises conserved regions complement Receptor type 1 (CR1) and CR2. GATAD2A gene is mapped to human chromosome 19p13.11.

Immunogen

Transcriptional repressor p66 α recombinant protein epitope signature tag (PrEST)

Application

Anti-GATAD2A antibody produced in rabbit has been used in western blotting[1] at 1:10000 dilution.
Anti-GATAD2A antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting.[1] To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

GATA zinc finger domain containing 2A (GATAD2A), a 68kDa protein, is component of a multi-subunit protein complex Mi-2/NuRD (Nucleosome Remodeling Deacetylase) complex. The Mi-2/NuRD complex is essential for the ATP-dependent chromatin remodeling and histone deacetylase activities. It also performs in the methylation dependent transcriptional repression by guiding repressors to the chromatin. It interacts with the histone tail in vitro and acetylates histone tails.
GATAD2A plays a key role in early mouse development. Deletion of GATAD2A leads to disruption of methyl-CpG binding domain 3 (Mbd3)/NuRD repression in in vivo studies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70974

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Bernaudo et al.
Journal of proteome research, 14(4), 1888-1899 (2015-03-17)
The study of protein-protein interactions is increasingly relying on mass spectrometry (MS). The classical approach of separating immunoprecipitated proteins by SDS-PAGE followed by in-gel digestion is long and labor-intensive. Besides, it is difficult to integrate it with most quantitative MS-based
Zongping Wang et al.
Oncology reports, 37(4), 2147-2152 (2017-03-06)
GATAD2A (GATA zinc finger domain containing 2A), is a subunit of NuRP (nucleosome remodeling and histone deacetylation) which plays key roles in tumor growth inhibition and embryonic development. However, its role in thyroid cancer remains unclear. In our study, we
Takahisa Kawaguchi et al.
PloS one, 13(1), e0185490-e0185490 (2018-02-01)
The genetic factors affecting the natural history of nonalcoholic fatty liver disease (NAFLD), including the development of nonalcoholic steatohepatitis (NASH) and NASH-derived hepatocellular carcinoma (NASH-HCC), are still unknown. In the current study, we sought to identify genetic factors related to
Y Xue et al.
Molecular cell, 2(6), 851-861 (1999-01-14)
ATP-dependent chromatin-remodeling complexes are known to facilitate transcriptional activation by opening chromatin structures. We report a novel human complex, named NURD, which contains not only ATP-dependent nucleosome disruption activity, but also histone deacetylase activity, which usually associates with transcriptional repression.
Marc Brackertz et al.
Nucleic acids research, 34(2), 397-406 (2006-01-18)
The Mi-2/NuRD complex is a multi-subunit protein complex with enzymatic activities involving chromatin remodeling and histone deacetylation. Targeting of Mi-2/NuRD to methylated CpG sequences mediates gene repression. The function of p66alpha and of p66beta within the multiple subunits has not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service