Skip to Content
Merck
All Photos(5)

Key Documents

HPA006313

Sigma-Aldrich

Anti-NR2C2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Orphan nuclear receptor TAK1 antibody produced in rabbit, Anti-Orphan nuclear receptor TR4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DGARQTGLLDPGMLVNIQQPLIREDGTVLLATDSKAETSQGALGTLANVVTSLANLSESLNNGDTSEIQPEDQSASEITRAFDTLAKALNTTDSSSSPSLADGIDTSGGGSIHVISRDQSTPIIEVEGPLLSDTHVTFKLTMPSPMP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR2C2(7182)

General description

NR2C2 (nuclear receptor subfamily 2, group C, member 2) is a protein encoded by the NR2C2 gene in humans and is mapped to chromosome to 3p25. It is a novel member of the nuclear hormone receptor family. It is localized to the hippocampus and the cerebellum in the granule cells. It may also have some role in the transcriptional regulation of neurons. It is found to be highly conserved in rat and human.

Immunogen

Orphan nuclear receptor TR4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Testicular nuclear receptor 4 (TR4) confers protection against oxidative stress and DNA damage, and may be involved in aging. It might have potential as a biomarker for prostate cancer (PCa) metastasis. Its interaction with keto-Mycolic acid (MA) is unique in that a host nuclear receptor binding with a bacterial lipid. It is involved in the final stages of differentiation of human erythroid cells, where it controls the basic and essential biological processes. It activates NF-κB in a genotoxic-stress mediated manner. Inactivation of this gene enhances apoptosis of neuroblastoma cells during chemotherapy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70756

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hedwin Kitdorlang Dkhar et al.
Journal of immunology (Baltimore, Md. : 1950), 193(1), 295-305 (2014-06-08)
The cell wall of Mycobacterium tuberculosis is configured of bioactive lipid classes that are essential for virulence and potentially involved in the formation of foamy macrophages (FMs) and granulomas. Our recent work established crosstalk between M. tuberculosis cell wall lipids
T Yoshikawa et al.
Genomics, 35(2), 361-366 (1996-07-15)
TR4 is a new member of the nuclear hormone receptor family. This receptor is highly conserved in rat and human, but an in-frame insertion of 19 amino acid residues in the amino-terminal (A/B) region was found in the human homolog
C Chang et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(13), 6040-6044 (1994-06-21)
We have identified a member of the steroid receptor superfamily and cloned it from human and rat hypothalamus, prostate, and testis cDNA libraries. The open reading frame between first ATG and terminator TGA can encode 615 (human) and 596 (rat)
Lihong Shi et al.
PLoS genetics, 10(5), e1004339-e1004339 (2014-05-09)
We previously reported that TR2 and TR4 orphan nuclear receptors bind to direct repeat (DR) elements in the ε- and γ-globin promoters, and act as molecular anchors for the recruitment of epigenetic corepressors of the multifaceted DRED complex, thereby leading
Yihui Fan et al.
Apoptosis : an international journal on programmed cell death, 18(10), 1224-1234 (2013-05-24)
Treatment failure in high risk neuroblastoma is largely due to development of chemoresistance. NF-κB activation is one of the resistance mechanisms for cancer cells to escape from chemotherapy-induced cell-death. TAK1 is an essential component in genotoxic stresses-induced NF-κB activation; however

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service