Skip to Content
Merck
All Photos(2)

Key Documents

HPA005908

Sigma-Aldrich

Anti-UCHL5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-UCH-L5, Anti-Ubiquitin C-terminal hydrolase UCH37, Anti-Ubiquitin carboxyl-terminal hydrolase isozyme L5, Anti-Ubiquitin thioesterase L5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

CTHQDVHLGETLSEFKEFSQSFDAAMKGLALSNSDVIRQVHNSFARQQMFEFDTKT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UCHL5(51377)

General description

Ubiquitin C-terminal hydrolase L5 (UCHL5), a cysteine protease,[1] is an integral part of the protein homeostasis network.[2] It belongs to the family of ubiquitin C-terminal hydrolases (UCHs).[1] UCHL5 is a proteasome-associated deubiquitinating enzyme[1], that is ubiquitously expressed in most of the normal human tissues.[2] UCHL5 gene is located on human chromosome 1q31.2.[2]

Immunogen

Ubiquitin carboxyl-terminal hydrolase isozyme L5 recombinant protein epitope signature tag (PrEST)

Application

Anti-UCHL5 antibody produced in rabbit has been used in immunohistochemistry(1:800).[1]

Biochem/physiol Actions

Ubiquitin C-terminal hydrolase L5 (UCHL5) is considered a new prognostic marker in rectal cancer and pancreatic ductal adenocarcinoma.[1] UCHL5 along with Rpn13 plays a key role in maintaining the progression of cell-cycle and DNA replication. It is necessary for effective polyubiquitin removal from proteasomal substrates, which leads to proteasomal substrate breakdown. The absence of UCHL5 may also inhibit the degradation of specific substrates, resulting in apoptosis.[2]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70227

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leena Arpalahti et al.
PloS one, 13(2), e0193125-e0193125 (2018-02-24)
Gastric cancer is the second most common cause of cancer-related mortality worldwide. Accurate prediction of disease progression is difficult, and new biomarkers for clinical use are essential. Recently, we reported that the proteasome-associated deubiquitinating enzyme UCHL5/Uch37 is a new prognostic
Shiho Fukui et al.
Oncotarget, 10(57), 5932-5948 (2019-11-02)
The ubiquitin-proteasome pathway plays an important role in the regulation of cellular proteins. As an alternative to the proteasome itself, recent research has focused on methods to modulate the regulation of deubiquitinating enzymes (DUBs) upstream of the proteasome, identifying DUBs
Leena Arpalahti et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 39(7), 1010428317716078-1010428317716078 (2017-07-07)
Colorectal cancer is among the three most common cancer types for both genders, with a rising global incidence. To date, prognostic evaluation is difficult and largely dependent on early detection and successful surgery. UCHL5/Uch37 is an integral part of the
Leena Arpalahti et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 39(6), 1010428317710411-1010428317710411 (2017-06-28)
Pancreatic ductal adenocarcinoma is a lethal disease with an overall 5-year survival of less than 5%. Prognosis among surgically treated patients is difficult and identification of new biomarkers is essential for accurate prediction of patient outcome. As part of one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service