Skip to Content
Merck
All Photos(2)

Documents

HPA005856

Sigma-Aldrich

Anti-TP53INP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DKFZp434M1317, Anti-FLJ10407, Anti-FLJ22139, Anti-NET3, Anti-P53DINP1, Anti-SIP, Anti-TP53INP1A, Anti-TP53INP1B, Anti-Teap, Anti-tumor protein p53 inducible nuclear protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VEAQNEMGQHIHCYVAALAAHTTFLEQPKSFRPSQWIKEHSERQPLNRNSLRRQNLTRDCHPRQVKHNGWVVHQPC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

tumor protein p53 inducible nuclear protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Tumor protein p53-inducible nuclear protein 1 is a protein encoded by the TP53INP1 gene in humans and is mapped to chromosome 8q22. It is referred as SIP, Teap, p53DINP1, TP53DINP1, TP53INP1A and TP53INP1B. It is a pro-apoptotic protein involved in cell stress response. Over-expression of this gene in numerous tissues leads to stress agents while down-expression is observed in stomach, pancreatic and inflammation-mediated colic carcinomas. Over-expression induces G1 cell cycle arrest and increases p53-mediated apoptosis. It may help in regulating p53-dependent apoptosis through phosphorylation of p53 at Ser46, serving as a cofactor for the putative p53-Ser46 kinase. It may play a significant role in the progression of anaplastic carcinoma. This gene may be considered as a relevant-specific target for molecular therapy of castration-resistant prostate cancer (CRPC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70321

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment of tumor protein p53 induced nuclear protein 1 (TP53INP1) gene to human chromosome band 8q22 by in situ hybridization.
J Nowak et al.
Cytogenetic and genome research, 97(1-2), 140E-140E (2002-11-20)
S Okamura et al.
Molecular cell, 8(1), 85-94 (2001-08-21)
Using the differential display method combined with a cell line that carries a well-controlled expression system for wild-type p53, we isolated a p53-inducible gene, termed p53DINP1 (p53-dependent damage-inducible nuclear protein 1). Cell death induced by DNA double-strand breaks (DSBs), as
Sophie Giusiano et al.
The Prostate, 72(12), 1286-1294 (2012-01-04)
Prostate cancer (PC) is one of the most common malignancies in industrialized countries, and the second leading cause of cancer-related death in the United States. We recently showed that over-expression of tumor protein 53-induced nuclear protein 1 (TP53INP1), a cell
Sophie Giusiano et al.
The Prostate, 72(2), 117-128 (2011-05-04)
Tumor protein 53-induced nuclear protein 1 (TP53INP1) is a proapoptotic protein involved in cell stress response. Whereas there is an overexpression of TP53INP1 in numerous tissues submitted to stress agents, TP53INP1 is down-expressed in stomach, pancreatic, and inflammation-mediated colic carcinomas.
Yasuhiro Ito et al.
Pathology, 38(6), 545-547 (2007-03-31)
Tumour protein p53-induced nuclear protein 1 (TP531NP1) is a stress-induced protein and plays a role in cell cycle arrest and p53-mediated apoptosis. In this study, we investigated TP531NP1 expression in human thyroid neoplasms. We immunohistochemically investigated TP531NP1 in 197 cases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service