Skip to Content
Merck
All Photos(10)

Key Documents

HPA005798

Sigma-Aldrich

Anti-GRHL1 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Grainyhead-like protein 1 homolog antibody produced in rabbit, Anti-Mammalian grainyhead antibody produced in rabbit, Anti-NH32 antibody produced in rabbit, Anti-Transcription factor CP2-like 2 antibody produced in rabbit, Anti-Transcription factor LBP-32 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TVFKPFIDLDTQPVLFIPDVHFANLQRGTHVLPIASEELEGEGSVLKRGPYGTEDDFAVPPSTKLARIEEPKRVLLYVRKESEEVFDALMLKTPSLKGL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRHL1(29841)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Grainyhead-like protein 1 homolog recombinant protein epitope signature tag (PrEST)

Application

Anti-GRHL1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Grainyhead-like 1 (GRHL1) is referred as MGR, NH32, LBP32 and TFCP2L2. This gene is critical for Drosophila neural development. Its overexpression in primary neuroblastomas is correlated on transcriptional and translational levels with favorable patient survival. It is an important regulator of the Dsg1 genes in the context of hair anchorage and epidermal differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70917

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johannes Fabian et al.
Cancer research, 74(9), 2604-2616 (2014-01-15)
Neuroblastoma is an embryonic solid tumor of neural crest origin and accounts for 11% of all cancer-related deaths in children. Novel therapeutic strategies are therefore urgently required. MYCN oncogene amplification, which occurs in 20% of neuroblastomas, is a hallmark of
Yiming He et al.
Cell death & disease, 12(5), 430-430 (2021-05-02)
Grainyhead-like 1 (GRHL1) is a transcription factor involved in embryonic development. However, little is known about the biological functions of GRHL1 in cancer. In this study, we found that GRHL1 was upregulated in non-small cell lung cancer (NSCLC) and correlated
Agnieszka Kikulska et al.
BMC cancer, 18(1), 23-23 (2018-01-06)
The Grainyhead-like (GRHL) transcription factors have been linked to many different types of cancer. However, no previous study has attempted to investigate potential correlations in expression of different GRHL genes in this context. Furthermore, there is very little information concerning
Kaiyu Kubota et al.
Molecular and cellular biology, 35(18), 3163-3177 (2015-07-08)
Placentation is a process that establishes the maternal-fetal interface and is required for successful pregnancy. The epithelial component of the placenta consists of trophoblast cells, which possess the capacity for multilineage differentiation and are responsible for placenta-specific functions. FOS-like antigen
Tomasz Wilanowski et al.
The EMBO journal, 27(6), 886-897 (2008-02-22)
In Drosophila, the grainy head (grh) gene plays a range of key developmental roles through the regulation of members of the cadherin gene family. We now report that mice lacking the grh homologue grainy head-like 1 (Grhl1) exhibit hair and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service