Skip to Content
Merck
All Photos(7)

Documents

HPA005741

Sigma-Aldrich

Anti-SELENBP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-56 kDa selenium-binding protein, Anti-SP56, Anti-Selenium-binding protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

IEPKDIHAKCELAFLHTSHCLASGEVMISSLGDVKGNGKGGFVLLDGETFEVKGTWERPGGAAPLGYDFWYQPRHNVMISTEWAAPNVLRDGFNPADV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SELENBP1(8991)

Immunogen

Selenium-binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Selenium-binding protein 1 is a protein encoded by the SELENBP1 gene in humans and is located on chromosome 1q21-22. It is referred as LPSB, SP56, hSBP, SBP56 and HEL-S-134P. It plays a critical role in prostate cells and is identified as a candidate anti-oncogene product. It may act as a potential pharmacological target for CRC treatment. Its expression is regulated by estrogen and its expression level may act as an important marker for predicting survival and effectiveness of selenium supplementation in breast cancer. It might also serve as a novel potential biomarker for early detection of squamous cell cancer (LSCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70248

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jee-Yeong Jeong et al.
BMB reports, 47(7), 411-416 (2014-05-31)
In the present study, we demonstrate that ectopic expression of 56-kDa human selenium binding protein-1 (hSP56) in PC-3 cells that do not normally express hSP56 results in a marked inhibition of cell growth in vitro and in vivo. Down-regulation of
Sheng Zhang et al.
PloS one, 8(5), e63702-e63702 (2013-05-25)
Supplemental dietary selenium is associated with reduced incidence of many cancers. The antitumor function of selenium is thought to be mediated through selenium-binding protein 1 (SELENBP1). However, the significance of SELENBP1 expression in breast cancer is still largely unknown. A
P W Chang et al.
Journal of cellular biochemistry, 64(2), 217-224 (1997-02-01)
We have isolated the full-length human 56 kDa selenium binding protein (hSP56) cDNA clone, which is the human homolog of mouse 56 kDa selenium binding protein. The cDNA is 1,668 bp long and has an open reading frame encoding 472
Gu-Qing Zeng et al.
PloS one, 8(8), e71865-e71865 (2013-08-27)
Our quantitative proteomic study showed that selenium-binding protein 1 (SELENBP1) was progressively decreased in human bronchial epithelial carcinogenic process. However, there is little information on expression and function of SELENBP1 during human lung squamous cell cancer (LSCC) carcinogenesis. iTRAQ-tagging combined
Ning Wang et al.
Oncology reports, 31(6), 2506-2514 (2014-04-17)
The aim of the present study was to examine the regulation of selenium binding protein 1 (SELENBP1) expression in colorectal cancer (CRC). Samples of cancer tissue and adjacent normal mucosa were collected from 83 CRC patients, and analyzed for SELENBP1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service