Skip to Content
Merck
All Photos(6)

Key Documents

HPA005469

Sigma-Aldrich

Anti-ANXA10 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Annexin A10, Anti-Annexin-10, Anti-Annexin-14

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

PPLYDAHELWHAMKGVGTDENCLIEILASRTNGEIFQMREAYCLQYSNNLQEDIYSETSGHFRDTLMNLVQGTREEGYTDPAMAAQDAMVLWEACQQKTGEHKTMLQMILCNK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA10(11199)

General description

Annexin A10 (ANXA10) belongs to the annexin A family, which binds to phospholipids in a calcium-regulated manner. Annexins generally contain a Ca2+-binding region which forms the C-terminal core domain, made of annexin repeats. The N-terminal domain is the ligand binding site and is hence, unique to each member. ANXA10 is normally expressed in the epithelia of the gastrointestinal tract. In humans, this gene is located on chromosome 4q33.

Immunogen

Annexin A10 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Annexin A (ANXA) family is involved in multiple biological functions such as cell growth, apoptosis, cell division and vesicle transport. The exact function of ANXA10 is still unknown. Studies suggest that ANXA10 is a potential marker for serrated adenocarcinoma (SAC). It can be used as a marker to differentiate between sporadic and LS-associated microsatellite-unstable colon cancer, as it is highly expressed in the former in comparison to the latter. This protein might have tumor suppressor function, as its down-regulation is associated with dedifferentiation, tumor progression and invasion. Low expression and homozygous deletion of ANXA10 is linked with poor prognosis in gastric and bladder cancer. Abnormal expression of this protein, along with that of osteopontin and α-fetoprotein, contributes to the progression and poor prognosis of hepatocellular carcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70304

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reetesh K Pai et al.
The American journal of surgical pathology, 38(4), 518-525 (2014-03-15)
Differentiating sporadic microsatellite-unstable colorectal carcinoma due to MLH1 promoter hypermethylation from Lynch syndrome (LS)-associated tumors due to mutations in mismatch-repair proteins is time consuming, cost intensive, and requires advanced laboratory testing. A mutation in BRAF has been shown to be
Xiaohui Liu et al.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban, 32(5), 669-674 (2012-10-18)
The effects of over-expression of ANXA10 gene on proliferation and apoptosis of hepato-cellular carcinoma cell line HepG2 were elucidated. The human ANXA10 gene was subcloned into the lentiviral vector, PGC-FU, to generate the lentiviral expression vector, PGC-FU-ANXA10. The corrected ANXA10
Nina Quiskamp et al.
Cellular and molecular life sciences : CMLS, 71(2), 311-329 (2013-05-30)
Annexin A10 is the latest identified member of the annexin family of Ca(2+)- and phospholipid-binding proteins. In previous studies, downregulation of annexin A10 was correlated with dedifferentiation, invasion, and tumor progression, pointing to a possible tumor suppressor role. However, the
R O Morgan et al.
Genomics, 60(1), 40-49 (1999-08-25)
We have identified and characterized a 12th subfamily of vertebrate annexins by systematic analysis of the primary structure, chromosomal mapping, and molecular evolution of unique cDNA and protein sequences from human and mouse. Distinctive features included rare expression, a codon
Sara A Sajanti et al.
Virchows Archiv : an international journal of pathology, 466(1), 5-12 (2014-11-15)
Serrated adenocarcinoma (SAC), representing at least 10 % of colorectal carcinomas (CRC), differs from conventional carcinomas not only by its histology, but also by its molecular basis. However, the diagnosis of SAC in poorly differentiated cases and without an adjacent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service