Skip to Content
Merck
All Photos(7)

Key Documents

HPA005466

Sigma-Aldrich

Anti-MYBBP1A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ37886, Anti-MYB binding protein (P160) 1a, Anti-P160, Anti-PAP2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EDLPLCSILQQIQEKYDLHQVKKAMLRPALFANLFGVLALFQSGRLVKDQEALMKSVKLLQALAQYQNHLQEQPRKALVDILSEVSKATLQEILPEVLKADLNIILSSPEQLELFLLAQQKVPSKLKKLVG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYBBP1A(10514)

General description

Myb-binding protein 1a (MYBBP1A) is a transcriptional co-regulator, which was originally identified as the protein which interacts with the proto-oncogene c-Myb. It is largely localized to the nucleolus, as it contains basic amino-acid repeats at its C-terminal. This protein weighs 160kDa and is expressed ubiquitously. Processing of the C-terminal nucleolar signal peptide produces two N-terminal forms of 140kDa and 67kDa. These forms are found in the nucleoplasm. MYBBP1A is located in the chromosomal region 17p13.3.

Immunogen

MYB binding protein (P160) 1a recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Myb-binding protein 1a (MYBBP1A) regulates essential functions such as cell division, proliferation and apoptosis. It binds to NFκB via its RelA/p65 subunit, and prevents it trans-activating capacity. It binds to multiple transcription factors, such as PPARγ co-activator 1α, and inhibits them. This protein also forms a part of the Ret-CoR co-repressor complex. It activates the aryl hydrocarbon receptor (AhR). It induces apoptosis by preventing Mdm2 mediated degradation of p53. MYBBP1A maintains the silent state of rDNA and negatively regulates the epigenetic state of silent rDNA. It also negatively regulates the expression of rRNA. MYBBP1A prevents breast cancer tumorigenesis and controls the metastasis as well as proliferation of head and neck squamous cell carcinoma cells.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86952

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuro Komatsu et al.
Traffic (Copenhagen, Denmark), 17(11), 1168-1180 (2016-10-27)
In adenoviral virions, the genome is organized into a chromatin-like structure by viral basic core proteins. Consequently viral DNAs must be replicated, chromatinized and packed into progeny virions in infected cells. Although viral DNA replication centers can be visualized by
R Keough et al.
Genomics, 62(3), 483-489 (2000-01-25)
We have previously isolated and characterized murine MYB binding protein (p160) 1a, a protein that specifically interacts with the leucine zipper motif within the negative regulatory domain of the c-Myb proto-oncoprotein. We now describe the molecular cloning of the human
Kensuke Akaogi et al.
BMC cancer, 13, 65-65 (2013-02-08)
Tumor suppressor p53 is mutated in a wide variety of human cancers and plays a critical role in anoikis, which is essential for preventing tumorigenesis. Recently, we found that a nucleolar protein, Myb-binding protein 1a (MYBBP1A), was involved in p53
Gustavo A Acuña Sanhueza et al.
BMC cancer, 12, 72-72 (2012-02-22)
Head and neck squamous cell carcinoma (HNSCC) is one of the most prevalent and lethal cancers worldwide and mortality mostly results from loco-regional recurrence and metastasis. Despite its significance, our knowledge on molecular, cellular and environmental mechanisms that drive disease
Tomohiro Yamauchi et al.
Genes to cells : devoted to molecular & cellular mechanisms, 13(1), 27-39 (2008-01-05)
Myb-binding protein 1a (Mybbp1a) was originally identified as a c-myb proto-oncogene product (c-Myb)-interacting protein, and also binds to various other transcription factors. The 160-kDa Mybbp1a protein (p160(MBP)) is ubiquitously expressed and is post-translationally processed in some types of cells to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service