Skip to Content
Merck
All Photos(5)

Key Documents

HPA005464

Sigma-Aldrich

Anti-FMNL2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Formin homology 2 domain-containing protein 2, Anti-Formin-like protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ERVEELEENISHLSEKLQDTENEAMSKIVELEKQLMQRNKELDVVREIYKDANTQVHTLRKMVKEKEEAIQRQSTLEKKIHELEKQGTIKIQKKGDGDIAILPVVASGTLSMGSEVVAGNSVGP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FMNL2(114793)

General description

Formin-like 2 (FMNL2) belongs to the family of diaphanous-related formins (DRF), which are ubiquitously expressed and have highly conserved domains. FMNL2 is highly expressed in mammary and gastrointestinal epithelia, placenta, reproductive tract and lymphatic tissues. FMNL2 has two isoforms due to alternative splicing, and they differ at their C-terminals. FMNL2A and FMNL2B have the characteristic FDD and formin homology1 (FH1) and FH2 domains. This gene is localized to chromosome 2q23.3.

Immunogen

Formin-like protein 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-FMNL2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Formin-like 2 (FMNL2) regulates actin cytoskeleton, and therefore controls cell motility and migration, cell adhesion, cell polarity, filopodium formation and cytokinesis. It mediates the polymerization of actin via its FH2 domain, while its FH1 domain binds to profilin. Cellular morphological changes are induced by FMNL2, via the N-myristoylation of the involved proteins. It also acts as the effector protein for GTPases belonging to Rho signaling pathway. FMNL2 is down-regulated in hepatocellular carcinoma as compared to normal hepatic epithelial cells. The lower expression of FMNL2 predicts poor prognosis and survival for hepatocellular carcinoma patients. In colorectal carcinoma, it has been implicated in maintaining the epithelial-mesenchymal transition (EMT). Studies show that this protein plays a key role in the metastasis of colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70050

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Grikscheit et al.
The Journal of cell biology, 209(3), 367-376 (2015-05-13)
Epithelial integrity is vitally important, and its deregulation causes early stage cancer. De novo formation of an adherens junction (AJ) between single epithelial cells requires coordinated, spatial actin dynamics, but the mechanisms steering nascent actin polymerization for cell-cell adhesion initiation
Hanna Grobe et al.
PloS one, 13(3), e0194716-e0194716 (2018-03-27)
De novo formation of epithelial cell-cell contacts relies on actin-based protrusions as well as tightly controlled turnover of junctional actin once cells encounter each other and adhesion complexes assemble. The specific contributions of individual actin regulators on either protrusion formation
Maria Gardberg et al.
BMC cell biology, 11, 55-55 (2010-07-17)
Diaphanous-related formins govern actin-based processes involved in many cellular functions, such as cell movement and invasion. Possible connections to developmental processes and cellular changes associated with malignant phenotype make them interesting study targets. In spite of this, very little is
Koko Moriya et al.
Bioscience, biotechnology, and biochemistry, 76(6), 1201-1209 (2012-07-14)
The subcellular localization of 13 recently identified N-myristoylated proteins and the effects of overexpression of these proteins on cellular morphology were examined with the aim of understanding the physiological roles of the protein N-myristoylation that occurs on these proteins. Immunofluorescence
Li Liang et al.
Human pathology, 42(11), 1603-1612 (2011-04-19)
The formin-like 2 protein is a member of the diaphanous-related formin family that controls actin-dependent processes such as cell motility and invasion. This study aimed at clarifying formin-like 2 expression in hepatocellular carcinoma and its correlation with clinicopathologic features and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service