Skip to Content
Merck
All Photos(2)

Key Documents

HPA005443

Sigma-Aldrich

Anti-CHI3L2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Chitinase 3-like protein 2 precursor antibody produced in rabbit, Anti-Chondrocyte protein 39 antibody produced in rabbit, Anti-YKL-39 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

IDPFLCSHLIYSFASIENNKVIIKDKSEVMLYQTINSLKTKNPKLKILLSIGGYLFGSKGFHPMVDSSTSRLEFINSIILFLRNHNFDGLDVSWIYPDQKENTHFTVLIHELAEAFQKDFTKSTKER

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHI3L2(1117)

General description

Chitinase 3-like 2 (CHI3L2) is very similar to CHI3L1, which in turn belongs to Chitinase-like protein (CLP) family. They have the same residues at their amino acid terminal. CHI3L2 is also called YKL39, and has a molecular weight of 39kDa and is produced and secreted by synoviocytes and articular chondrocytes. It was first obtained from the primary culture of human articular cartilage chondrocytes, where it co-purified with CHI3L1.

Immunogen

Chitinase 3-like protein 2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Unlike other CLPs (chitinase-like proteins), Chitinase 3-like 2 (CHI3L2), has no hydrolase activity and therefore functions as a pseudo-chitinase. However, it binds to chitooligosaccharides and retains the chitinase-like ligand binding characteristics. Studies suggest that CHI3L2 might act as a growth and differentiation factor in cartilage homeostasis. It might also aid the progression of osteoarthritis as its expression is increased in osteoarthritic articular chondrocytes. It regulates proliferation and differentiation in human embryonic kidney (HEK293) and human glioblastoma (U87 MG) cells via the extracellular signal-regulated kinase (ERK) signaling pathway. High levels of CHI3L2 mRNA have been detected in lung and heart and its expression is up regulated in glioblastomas.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70015

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Areshkov et al.
TSitologiia i genetika, 44(1), 3-9 (2010-03-06)
Human cartilage chitinase 3-like protein 2 (CHI3L2, YKL-39) is secreted by articular chondrocytes, also synoviocytes, lung, and heart. Increased levels of YKL-39 have been demonstrated in synovial fluids of patients with rheumatoid or osteoarthritis as well as in some other
Kazumasa Miyatake et al.
Biochemical and biophysical research communications, 431(1), 52-57 (2013-01-08)
Human YKL39 (chitinase 3-like protein 2/CHI3L2) is a secreted 39kDa protein produced by articular chondrocytes and synoviocytes. Recent studies showed that hYKL-39 expression is increased in osteoarthritic articular chondrocytes suggesting the involvement of hYKL-39 in the progression of osteoarthritis (OA).
Marianne Schimpl et al.
The Biochemical journal, 446(1), 149-157 (2012-06-30)
The chitinase-like proteins YKL-39 (chitinase 3-like-2) and YKL-40 (chitinase 3-like-1) are highly expressed in a number of human cells independent of their origin (mesenchymal, epithelial or haemapoietic). Elevated serum levels of YKL-40 have been associated with a negative outcome in
Pavlo O Areshkov et al.
International journal of biological sciences, 8(1), 39-48 (2012-01-03)
The activation of extracellular signal-regulated kinases (ERK1/2) has been associated with specific outcomes. Sustained activation of ERK1/2 by nerve growth factor (NGF) is associated with translocation of ERKs to the nucleus of PC12 cells and precedes their differentiation into sympathetic-like

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service