Skip to Content
Merck
All Photos(4)

Key Documents

HPA004911

Sigma-Aldrich

ANTI-HNRNPD antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AU-rich element RNA-binding protein 1, Anti-HNRPD, Anti-Heterogeneous nuclear ribonucleoprotein D0, Anti-hnRNP D0

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

RGFCFITFKEEEPVKKIMEKKYHNVGLSKCEIKVAMSKEQYQQQQQWGSRGGFAGRARGRGGDQQSGYGKVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HNRNPD(3184)

General description

HNRNPD (heterogeneous nuclear ribonucleoprotein D) is a rhythmical RNA-binding protein. It consists of two ribonucleoprotein (RNP) type RNA-binding domains (RBDs) that are responsible for binding of UUAG sequence.

Immunogen

Heterogeneous nuclear ribonucleoprotein D0 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

HNRNPD (heterogeneous nuclear ribonucleoprotein D) is involved in the cryptochrome 1 (Cry1) mRNA degradation and rhythmic translation. In addition to translational control, it also contributes to the cell cycle, apoptosis, and inflammation. HNRNPD directly interacts with the 3′ untranslated region (UTR) of Cry1 for the translational regulation of Cry1 mRNA. This interaction is a vital step for rhythmic translation. After a series of interactions, HNRNPD recruits 40S ribosomal subunit to the 5′ end of mRNA to initiate the translation of Cry1 mRNA.[1] Its expression is up-regulated in several tumors. It has been reported that HNRNPD may have important role in identification and progression of the hepatocellular carcinoma (HCC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86960

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingzhuo Yang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(3), 2747-2751 (2013-11-12)
RNA-binding factor 1 (AUF1) was found to be up-regulated in numerous tumors compared with untransformed tissues. Furthermore, it has been identified to regulate mRNAs en masse in hepatocellular carcinoma (HCC). Metadherin (MTDH) as a novel oncogene also promotes tumor progression
Kyung-Ha Lee et al.
Nucleic acids research, 42(6), 3590-3606 (2014-01-16)
In the present study, we investigated the 3' untranslated region (UTR) of the mouse core clock gene cryptochrome 1 (Cry1) at the post-transcriptional level, particularly its translational regulation. Interestingly, the 3'UTR of Cry1 mRNA decreased its mRNA levels but increased
T Nagata et al.
Journal of molecular biology, 287(2), 221-237 (1999-03-19)
Heterogeneous nuclear ribonucleoprotein (hnRNP) D0 has two ribonucleoprotein (RNP)-type RNA-binding domains (RBDs), each of which can bind solely to the UUAG sequence specifically. The structure of the N-terminal RBD (RBD1) determined by NMR is presented here. It folds into a
Dominik Peters et al.
The Journal of biological chemistry, 292(20), 8331-8341 (2017-03-30)
The cyclin-dependent kinase inhibitor p21 is an important player in stress pathways exhibiting both tumor-suppressive and oncogenic functions. Thus, expression of p21 has to be tightly controlled, which is achieved by numerous mechanisms at the transcriptional, translational, and posttranslational level.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service