Skip to Content
Merck
All Photos(7)

Key Documents

HPA004141

Sigma-Aldrich

Anti-ACSS2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ACS antibody produced in rabbit, Anti-AceCS antibody produced in rabbit, Anti-Acetate-CoA ligase antibody produced in rabbit, Anti-Acetyl-CoA synthetase antibody produced in rabbit, Anti-Acetyl-coenzyme A synthetase, cytoplasmic antibody produced in rabbit, Anti-Acyl-CoA synthetase short-chain family member 2 antibody produced in rabbit, Anti-Acyl-activating enzyme antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Check Cart for Availability


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PGETTQITYHQLLVQVCQFSNVLRKQGIQKGDRVAIYMPMIPELVVAMLACARIGALHSIVFAGFSSESLCERILDSSCSLLITTDAFYRGEKLVNLKELADEALQKCQEKGFPVRCCIVVKHLGRAELGMGDSTSQSPPIKRSCPDVQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACSS2(55902)

Immunogen

Acetyl-coenzyme A synthetase, cytoplasmic recombinant protein epitope signature tag (PrEST)

Application

Anti-ACSS2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues.[1] These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ACSS2 encodes a cytosolic enzyme, acetyl-CoA synthetase (ACS). It is highly essential along with Hypoxia Inducible Factor 2 (HIF-2) for maximal colony formation, proliferation, migration, and invasion during stress. It stimulates the activation reaction of acetate for lipid synthesis and energy generation. Acetate plays a major role in almost all of the metabolic pathways. Monomeric form of the ACSS2 generates acetate in a ATP dependent reaction. The activity is controlled by sterol regulatory element-binding proteins (SREBPs). It is clinically associated with the cancer cell growth under low-oxygen and lipid-depleted conditions.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86744

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minghua Li et al.
Cell biology international, 45(6), 1278-1287 (2021-02-10)
One hallmark of renal cell carcinoma (RCC) is metabolic reprogramming, which involves elevation of glycolysis and upregulation of lipid metabolism. However, the mechanism of metabolic reprogramming is incompletely understood. Monocarboxylate transporter 1 (MCT1) promotes transport for lactate and pyruvate, which
A Luong et al.
The Journal of biological chemistry, 275(34), 26458-26466 (2000-06-14)
Through suppressive subtractive hybridization, we identified a new gene whose transcription is induced by sterol regulatory element-binding proteins (SREBPs). The gene encodes acetyl-CoA synthetase (ACS), the cytosolic enzyme that activates acetate so that it can be used for lipid synthesis
Zachary T Schug et al.
Cancer cell, 27(1), 57-71 (2015-01-15)
A functional genomics study revealed that the activity of acetyl-CoA synthetase 2 (ACSS2) contributes to cancer cell growth under low-oxygen and lipid-depleted conditions. Comparative metabolomics and lipidomics demonstrated that acetate is used as a nutritional source by cancer cells in
Rui Chen et al.
PloS one, 10(2), e0116515-e0116515 (2015-02-18)
Optimal stress signaling by Hypoxia Inducible Factor 2 (HIF-2) during low oxygen states or hypoxia requires coupled actions of a specific coactivator/lysine acetyltransferase, Creb binding protein (CBP), and a specific deacetylase, Sirtuin 1 (SIRT1). We recently reported that acetylation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service