Skip to Content
Merck
All Photos(7)

Key Documents

HPA003916

Sigma-Aldrich

Anti-SMARCE1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BRG1-associated factor 57, Anti-SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

EEREKEAAEQAERSQSSIVPEEEQAANKGEEKKDDENIPMETEETHLEETTESQQNGEEGTSTPEDKESGQEGVDSMAEEGTSDSNTGSESNSATVEEPPTDPIPEDEK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMARCE1(6605)

Immunogen

SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1 is a protein encoded by the SMARCE1 gene in humans. It is also referred to as BAF57. It is a component of the switching-defective and sucrose nonfermenting (SWI/SNF) chromatin-remodeling complex conglomerate that modulates androgen receptor activity to promote prostate cancer. It is involved in chromatin remodeling and is associated with pathogenesis of both meningiomas and tumors with clear-cell histology. Its activity may be one of the candidates for endometrial cancer therapy, especially therapy for aggressive tumors showing overexpression of p53. Mutation in this gene causes spinal clear cell meningiomas.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86684

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam J Smith et al.
The Journal of pathology, 234(4), 436-440 (2014-08-22)
We recently reported SMARCE1 mutations as a cause of spinal clear cell meningiomas. Here, we have identified five further cases with non-NF2 spinal meningiomas and six with non-NF2 cranial meningiomas. Three of the spinal cases and three of the cranial
Arnault Tauziede-Espariat et al.
Brain pathology (Zurich, Switzerland), 28(4), 466-474 (2017-05-06)
Clear cell meningioma (CCM) is a rare grade II histopathological subtype that usually occurs in young patients and displays high recurrence rate. Germline SMARCE1 mutations have been described in hereditary forms of this disease and more recently in small syndromic
Miriam J Smith et al.
Histopathology, 70(5), 814-820 (2016-11-29)
Clear cell meningioma (CCM) is a rare subtype of meningioma and shows not only unusual histology, but also unique clinical features. Recently, SMARCE1 mutations have been shown to cause spinal and cranial CCMs. We present 12 cases which were diagnosed
Sucharitha Balasubramaniam et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 19(10), 2657-2667 (2013-03-16)
BAF57, a component of the switching-defective and sucrose nonfermenting (SWI/SNF) chromatin-remodeling complex conglomerate, modulates androgen receptor activity to promote prostate cancer. However, the molecular consequences of tumor-associated BAF57 expression have remained undefined in advanced disease such as castration-resistant prostate cancer
Dorian Hirschmann et al.
Wiener klinische Wochenschrift (2024-05-31)
To identify factors for tumor relapse and poor outcome in patients with meningiomas in the first two decades of life. All patients ≤ 21 years of age who underwent resection of a meningioma at the department of neurosurgery, Medical University of Vienna between

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service