Skip to Content
Merck
All Photos(8)

Key Documents

HPA003624

Sigma-Aldrich

Anti-RBM3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IS1-RNPL, Anti-RNA binding motif (RNP1, RRM) protein 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€544.00


Estimated to ship on02 June 2025


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DEQALEDHFSSFGPISEVVVVKDRETQRSRGFGFITFTNPEHASVAMRAMNGESLDGRQIRVDHAGKSARGTRGGGFGAHGRGRSYSRGGGDQGYGSGRYYDSRPGGYGYGYGRSRDYNGRNQGGYDRYSGGNY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RBM3(5935)

General description

The RBM3 gene is mapped to human chromosome Xp11.23. The gene is a X-chromosome related gene that encodes an alternatively spliced transcript that contains a RNA recognition motif (RRM) domain and a glycine-rich region. This protein is capable of binding to both DNA and RNA.

Immunogen

RNA binding motif (RNP1, RRM) protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Putative RNA-binding protein 3 is a protein encoded by the RBM3 gene in humans. The RNA-binding proteins (RBPs) function in RNA metabolism and regulate gene transcription. RBM3 is transcriptionally induced by low temperature and hypoxia. It is found to be up-regulated in human tumors and its overexpression may serve as an important molecular mechanism for astrocytic carcinogenesis. The gene may have proliferative and/or proto-oncogenic functions in human astrocytomas. Loss of this gene expression is associated with aggressive tumours and an independent factor of poor prognosis in patients with urothelial bladder cancer.[1] Over expression of this gene is associated with good prognosis in breast cancer, ovarian cancer, malignant melanoma and colorectal cancer.[2]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74363

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Grupp et al.
BMC cancer, 18(1), 1106-1106 (2018-11-14)
RBM3 expression has been suggested as prognostic marker in several cancer types. The purpose of this study was to assess the prevalence and clinical significance of altered RBM3 expression in esophageal cancer. RBM3 protein expression was measured by immunohistochemistry using
Hai-Tao Zhang et al.
Chinese medical journal, 126(10), 1948-1952 (2013-05-16)
The RNA-binding motif protein 3 (RBM3), which is transcriptionally induced by low temperature and hypoxia, has recently been found to be upregulated in human tumors. However, its expression status in human astrocytoma is not well defned. This article focuses on
Liv Jonsson et al.
Diagnostic pathology, 6, 91-91 (2011-10-01)
High expression of the RNA-binding protein RBM3 has previously been found to be associated with good prognosis in breast cancer, ovarian cancer, malignant melanoma and colorectal cancer. The aim of this study was to examine the prognostic impact of immunohistochemical
Karolina Boman et al.
BMC urology, 13, 17-17 (2013-04-10)
Low nuclear expression of the RNA-binding motif protein 3 (RBM3) has previously been found to be associated with poor prognosis in several cancer forms e.g. breast, ovarian, colorectal, prostate cancer and malignant melanoma. The aim of this study was to
Nathaniel Melling et al.
Histopathology, 68(2), 191-198 (2015-04-30)
RNA-binding motif protein 3 (RBM3) has recently been suggested as a prognostic biomarker in an array of human cancers. This study aimed to examine its effects in colorectal cancers. RBM3 expression was analysed by immunohistochemistry on a tissue microarray containing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service