Skip to Content
Merck
All Photos(8)

Key Documents

HPA003509

Sigma-Aldrich

Anti-UBE2J1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HSUBC6e, Anti-NCUBE1, Anti-Non-canonical ubiquitin-conjugating enzyme 1, Anti-UBE2J1, Anti-Ubiquitin-conjugating enzyme E2 J1, Anti-Yeast ubiquitin-conjugating enzyme UBC6 homolog E

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VLLPLKSGSDSSQADQEAKELARQISFKAEVNSSGKTISESDLNHSFSLTDLQDDIPTTFQGATASTSYGLQNSSAASFHQPTQPVAKNTSMSPRQRRAQQQSQRRLSTSPDVIQGHQPRDNHTD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2J1(51465)

Immunogen

Ubiquitin-conjugating enzyme E2 J1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ubiquitin-conjugating enzyme E2 J1 is a protein encoded by the UBE2J1 gene in humans. In yeast, the enzyme is localized to the cytoplasmic surface of the ER and participates in ER-associated degradation (ERAD) of misfolded proteins. It possesses a transmembrane domain that anchors it to the cytoplasmic surface of the endoplasmic reticulum and is involved in targeting retrotranslocated, ER-associated proteins for proteasomal degradation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86439

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uwe Lenk et al.
Journal of cell science, 115(Pt 14), 3007-3014 (2002-06-26)
Integral membrane and secretory proteins which fail to fold productively are retained in the endoplasmic reticulum and targeted for degradation by cytoplasmic proteasomes. Genetic and biochemical analyses suggest that substrates of this pathway must be dislocated across the membrane of
D Lester et al.
Biochemical and biophysical research communications, 269(2), 474-480 (2000-03-10)
Ubiquitin-conjugating enzymes (UBCs) selectively target proteins for proteasomal degradation by the covalent attachment of ubiquitin moieties. Yeast UBC6 is unusual in having an active site distinct from all other UBCs and in possessing a transmembrane domain that anchors it to
Carla Rodriguez Tirado et al.
Oncogene, 43(4), 265-280 (2023-11-30)
Prostate cancer (PCa) is primarily driven by aberrant Androgen Receptor (AR) signaling. Although there has been substantial advancement in antiandrogen therapies, resistance to these treatments remains a significant obstacle, often marked by continuous or enhanced AR signaling in resistant tumors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service