Skip to Content
Merck
All Photos(6)

Key Documents

HPA003494

Sigma-Aldrich

Anti-LCK antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LSK, Anti-Lymphocyte cell-specific protein-tyrosine kinase, Anti-Proto-oncogene tyrosine-protein kinase LCK, Anti-T cell-specific protein-tyrosine kinase, Anti-p56-LCK

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€482.00

€482.00


Estimated to ship on15 April 2025



Select a Size

Change View
100 μL
€482.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€482.00


Estimated to ship on15 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MGCGCSSHPEDDWMENIDVCENCHYPIVPLDGKGTLLIRNGSEVRDPLVTYEGSNPPASPLQDNLVIALHSYEPSHDGDLGFEKGEQLRILEQSGEWWKAQSLTTGQEGFIPFNFVAKANSLEPEPWFFKNLSRKDAERQLLAPGNTHGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LCK(3932)

Immunogen

LCK proto-oncogene, Src family tyrosine kinase

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Lck (or lymphocyte-specific protein tyrosine kinase) is a 56kDa protein and is found inside specialized cells of the immune system called lymphocytes. It plays a crucial role in the T-cell response by transducing early activation signals triggered by TCR (T-cell receptor) engagement. These signals result in the phosphorylation of immunoreceptor tyrosine-based activation motifs that is present within the cytosolic tails of the TCR-associated CD3 subunits. After phosphorylation, they serve as scaffolds for the assembly of a large supramolecular signaling complex responsible for T-cell activation. This gene plays a major role in proximal and distal B-cell receptor (BCR) -mediated signaling in chronic lymphocytic leukemia (CLL) cells. The Lck conformational states represent an intrinsic mechanism for the intermolecular organization of early T cell signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78895

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leandro N Ventimiglia et al.
The Biochemical journal, 454(2), 169-179 (2013-08-13)
Tyrosine phosphorylation is one of the key covalent modifications that occur in multicellular organisms. Since its discovery more than 30 years ago, tyrosine phosphorylation has come to be understood as a fundamentally important mechanism of signal transduction and regulation in
Tomoko Miyata-Takata et al.
Histopathology, 73(6), 1030-1038 (2018-08-14)
Although the neoplastic cells of extranodal natural killer (NK)/T-cell lymphoma (ENKTL) usually do not express T-cell antigens, the T-cell receptor (TCR) gene might be rearranged and TCR protein expressed. The aim is to elucidate the expression of the downstream TCR
Fatima Talab et al.
Molecular cancer research : MCR, 11(5), 541-554 (2013-03-19)
B-cell receptor (BCR) signals promote survival of chronic lymphocytic leukemia (CLL) cells, and it is believed that overexpressed and constitutively active Lyn mediates this signaling. Here, we show that CLL cells express lymphocyte-specific protein tyrosine kinase (LCK) and that inhibition
Jérémie Rossy et al.
Nature immunology, 14(1), 82-89 (2012-12-04)
Phosphorylation of the T cell antigen receptor (TCR) by the tyrosine kinase Lck is an essential step in the activation of T cells. Because Lck is constitutively active, spatial organization may regulate TCR signaling. Here we found that Lck distributions
Louise A Stephen et al.
Developmental cell, 47(1), 122-132 (2018-09-18)
Upon engagement of the T cell receptor with an antigen-presenting cell, LCK initiates TCR signaling by phosphorylating its activation motifs. However, the mechanism of LCK activation specifically at the immune synapse is a major question. We show that phosphorylation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service