Skip to Content
Merck
All Photos(5)

Key Documents

HPA003432

Sigma-Aldrich

Anti-ITGA8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Integrin alpha-8 precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200-1:500

immunogen sequence

HKEEEVGPLVEHIYELHNIGPSTISDTILEVGWPFSARDEFLLYIFHIQTLGPLQCQPNPNINPQDIKPAASPEDTPELSAFLRNSTIPHLVRKRDVHVVEFHRQSPAKILNCTNIECLQISCA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGA8(8516)

General description

Integrin α-8 (ITGA8) is a transmembrane protein.[1] This protein is encoded by the ITGA8 gene in humans. The gene is highly expressed in smooth muscle and smooth muscle-like contractile cells. The ITGA8 gene is located on the human chromosome at 10p13.

Immunogen

Integrin alpha-8 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ITGA8 antibody produced in rabbit has been used in tissue labeling.[1]

Biochem/physiol Actions

Integrin α-8 (ITGA8) plays a role in cell adhesion and cell signaling.[1]
Integrin a-8 (ITGA8) plays an important role in organogenesis and is also expressed highly during kidney and lung development. It may be associated with schizophrenia. Mutation in this gene causes bilateral renal agenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85140

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camille Humbert et al.
American journal of human genetics, 94(2), 288-294 (2014-01-21)
Renal hypodysplasia (RHD) is a heterogeneous condition encompassing a spectrum of kidney development defects including renal agenesis, hypoplasia, and (cystic) dysplasia. Heterozygous mutations of several genes have been identified as genetic causes of RHD with various severity. However, these genes
Li-yi Cai et al.
Life sciences, 80(16), 1458-1465 (2007-02-17)
Methylated promoter CpG islands (CGIs) can be used to find novel tumor-suppressor genes and disease markers. In this study, to identify promoter CGIs aberrantly methylated in human ovarian cancers, we performed a genome-wide screening for differentially methylated DNA fragments using
Jared Coffin Talbot et al.
Developmental biology, 416(1), 136-148 (2016-06-07)
Both Fras1 and Itga8 connect mesenchymal cells to epithelia by way of an extracellular 'Fraser protein complex' that functions in signaling and adhesion; these proteins are vital to the development of several vertebrate organs. We previously found that zebrafish fras1
Catherine Ekwa-Ekoka et al.
Matrix biology : journal of the International Society for Matrix Biology, 23(7), 487-496 (2004-12-08)
The integrin alpha8 is highly expressed during kidney and lung development. alpha8-deficient mice display abnormal renal development suggesting that alpha8 plays a critical role in organogenesis. Therefore, it would be of considerable interest to understand the genomic structure, localization and
Irwan Supriyanto et al.
Progress in neuro-psychopharmacology & biological psychiatry, 40, 347-352 (2012-11-17)
Cell adhesion molecules (CAMs) play pivotal role in the development of the central nervous system (CNS) and have also been reported to play role in the pathophysiology of schizophrenia. Missense mutations in the CAMs genes might alter the binding of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service