Skip to Content
Merck
All Photos(1)

Documents

HPA003408

Sigma-Aldrich

Anti-IFIT2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IFI-54K antibody produced in rabbit, Anti-IFIT-2 antibody produced in rabbit, Anti-ISG-54 K antibody produced in rabbit, Anti-Interferon-induced 54 kDa protein antibody produced in rabbit, Anti-Interferon-induced protein with tetratricopeptide repeats 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

RVCSILASLHALADQYEDAEYYFQKEFSKELTPVAKQLLHLRYGNFQLYQMKCEDKAIHHFIEGVKINQKSREKEKMKDKLQKIAKMRLSKNGADSEALHVLAFLQELNEKMQQADEDSERGLESGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFIT2(3433)

Immunogen

Interferon-induced protein with tetratricopeptide repeats 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-IFIT2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Interferon-induced protein with tetratricopeptide repeats 2 or IFIT2 is a protein encoded by the IFIT2 gene in humans. It is one of the most highly responsive interferon-stimulated genes and is responsible for interferon stimulation. It may play an important role in inhibiting cell migration. The protein may help in preventing oral squamous cell carcinoma (OSCC) metastasis and may serve as a valuable prognostic marker. It negatively regulates eIF3 protein complex and inhibits translation initiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86653

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K C Lai et al.
Oncogene, 32(32), 3686-3697 (2012-09-19)
Interferon-induced protein with tetratricopeptide repeats 2 (IFIT2) is one of the most highly responsive interferon-stimulated genes, but its biological functions are poorly understood. In this study, we aimed to explore the underlying mechanisms by which depleting IFIT2 induces the migration
Ranran Wang et al.
Journal of virology, 97(4), e0190722-e0190722 (2023-03-23)
Merkel cell polyomavirus (MCPyV) has been associated with approximately 80% of Merkel cell carcinoma (MCC), an aggressive and increasingly incident skin cancer. The link between host innate immunity, viral load control, and carcinogenesis has been established but poorly characterized. We
Xiaoshan Feng et al.
BMC cancer, 14, 633-633 (2014-09-02)
An increasing body of evidence indicates that miRNAs have a critical role in carcinogenesis and cancer progression; however, the role of miRNAs in the tumorigenesis of adencarcinoma of gastric esophageal junction (AGEJ) remains largely unclear. The SGC7901 and BGC-823 gastric
Kuo-Chu Lai et al.
Molecular cancer research : MCR, 6(9), 1431-1439 (2008-09-30)
The function of the IFN-stimulated gene family protein, IFN-induced protein with tetratricopeptide repeats 2 (IFIT2), is poorly understood. Here, we report that IFIT2 colocalizes with cytokeratin 18 in oral squamous cell carcinoma (OSCC) cells. Treatment of OSCC cells with IFN-beta
Susanne Berchtold et al.
BMC immunology, 9, 75-75 (2008-12-26)
Interferon induced tetratricopeptide repeat protein 2 (IFIT-2, P54) belongs to the type I interferon response genes and is highly induced after stimulation with LPS. The biological function of this protein is so far unclear. Previous studies indicated that IFIT-2 binds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service