Skip to Content
Merck
All Photos(8)

Key Documents

HPA003279

Sigma-Aldrich

Anti-OPTN antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FIP-2, Anti-FIP2, Anti-GLC1E, Anti-HIP7, Anti-HYPL, Anti-NRP, Anti-TFIIIA-INTP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LELAEKALASKQLQMDEMKQTIAKQEEDLETMTILRAQMEVYCSDFHAERAAREKIHEEKEQLALQLAVLLKENDAFEDGGRQSLMEMQSRHGARTSDSDQQAYLVQRGAEDRDWRQQRNIPIHSCPKCGEVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OPTN(10133)

Looking for similar products? Visit Product Comparison Guide

General description

OPTN (optineurin) is a multifunctional 74kDa protein, which is composed of multiple coiled-coil domains, a leucine zipper, a ubiquitin-binding domain (UBD), and its C-terminus contains a C2H2 type zinc finger. It is expressed ubiquitously, and has predominant expression in brain, retina, heart, kidney, testis, skeletal muscle and placenta. It localizes to recycling endosomes, Golgi bodies and cytoplasm.[1] This gene is localized to human chromosome 10p13.

Immunogen

Optineurin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

OPTN (Optineurin) functions in several processes including vesicular trafficking from the Golgi to the plasma membrane, regulation of NF-κB, signal transduction and gene expression.[1] It mediates the linking of myosin VI to the Golgi complex and functions in the formation of Golgi ribbon and exocytosis. It negatively regulates induction of IFNβ involved in antiviral signaling and serves as a potential target for antiviral therapy.[2] It binds to Rab8 and regulates its association with TBC1D17, a Rab GTPase-activating protein, thus affecting Rab8-mediated processes, including the trafficking of transferrin receptor from the early endosome to the recycling endosome. Mutations in this gene cause primary open-angle glaucoma (POAG), a neurodegenerative eye disease that causes blindness.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85211

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vipul Vaibhava et al.
Journal of cell science, 125(Pt 21), 5026-5039 (2012-08-03)
Rab GTPases regulate various membrane trafficking pathways but the mechanisms by which GTPase-activating proteins recognise specific Rabs are not clear. Rab8 is involved in controlling several trafficking processes, including the trafficking of transferrin receptor from the early endosome to the
Laëtitia Michou et al.
Bone, 51(4), 720-728 (2012-07-17)
We performed a genetic association study of rare variants and single nucleotide polymorphisms (SNPs) of UCMA/GRP and OPTN genes, in French-Canadian patients with Paget's disease of bone (PDB) and in healthy controls from the same population. We reproduced the variant
Ananthamurthy Nagabhushana et al.
BMC cell biology, 11, 4-4 (2010-01-21)
Optineurin is a multifunctional protein involved in several functions such as vesicular trafficking from the Golgi to the plasma membrane, NF-kappaB regulation, signal transduction and gene expression. Mutations in optineurin are associated with glaucoma, a neurodegenerative eye disease that causes
Tayebeh Rezaie et al.
Science (New York, N.Y.), 295(5557), 1077-1079 (2002-02-09)
Primary open-angle glaucoma (POAG) affects 33 million individuals worldwide and is a leading cause of blindness. In a study of 54 families with autosomal dominantly inherited adult-onset POAG, we identified the causative gene on chromosome 10p14 and designated it OPTN
Daniela A Sahlender et al.
The Journal of cell biology, 169(2), 285-295 (2005-04-20)
Myosin VI plays a role in the maintenance of Golgi morphology and in exocytosis. In a yeast 2-hybrid screen we identified optineurin as a binding partner for myosin VI at the Golgi complex and confirmed this interaction in a range

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service