Skip to Content
Merck
All Photos(3)

Key Documents

HPA003221

Sigma-Aldrich

Anti-RPS6KA3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-90 kDa ribosomal protein S6 kinase 3, Anti-ISPK-1, Anti-Insulin-stimulated protein kinase 1, Anti-MAP kinase-activated protein kinase 1b, Anti-MAPKAPK1, Anti-RSK-2, Anti-Ribosomal S6 kinase 2, Anti-Ribosomal protein S6 kinase alpha-3, Anti-S6K-alpha 3, Anti-p90-RSK 3, Anti-pp90RSK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

MPLAQLADPWQKMAVESPSDSAENGQQIMDEPMGEEEINPQTEEVSIKEIAITHHVKEGHEKADPSQFELLKVLGQGSFGKVFLVKKISGSDARQLYAMKVLKKATLKVRDRVRTKMERDILVEVNHPFIVKLHYAFQTEGKLY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPS6KA3(6197)

General description

The RPS6KA3 (ribosomal protein S6 kinase, 90kDa, polypeptide 3) gene, also referred to as RSK2, is mapped to human chromosome Xp22.2. It encodes a protein belonging to the RSK (ribosomal S6 kinase) family of serine/threonine kinases.

Immunogen

Ribosomal protein S6 kinase alpha-3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RPS6KA3 (ribosomal protein S6 kinase, 90kDa, polypeptide 3) gene encodes a protein that may function in cell growth and differentiation. Integrin-regulated RSK signaling regulates cytokinesis in epithelial cells. It plays an important role as a signal integrator to provide antianoikis protection to cancer cells leading to cancer cell invasion and tumor metastasis. It is involved in mitogenic and stress-induced activation of certain transcription factors, cellular proliferation, survival, and differentiation. Mutations in this gene cause Coffin-Lowry syndrome that is characterized by severe psychomotor retardation, facial and digital dysmorphisms, and progressive skeletal deformations.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85204

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shomita S Mathew et al.
Journal of cell science, 127(Pt 3), 534-545 (2013-11-29)
Cytokinesis is the final stage in cell division. Although integrins can regulate cytokinesis, the mechanisms involved are not fully understood. In this study, we demonstrate that integrin-regulated ERK (extracellular signal-related kinase) and RSK (p90 ribosomal S6 kinase) signaling promotes successful
E Trivier et al.
Nature, 384(6609), 567-570 (1996-12-12)
The Coffin-Lowry syndrome (CLS), an X-linked disorder, is characterized by severe psychomotor retardation, facial and digital dysmorphisms, and progressive skeletal deformations. Genetic linkage analysis mapped the CLS locus to an interval of 2-3 megabases at Xp22.2. The gene coding for
Lingtao Jin et al.
Molecular and cellular biology, 33(13), 2574-2585 (2013-04-24)
How invasive and metastatic tumor cells evade anoikis induction remains unclear. We found that knockdown of RSK2 sensitizes diverse cancer cells to anoikis induction, which is mediated through phosphorylation targets including apoptosis signal-regulating kinase 1 (ASK1) and cyclic AMP (cAMP)
F Abidi et al.
European journal of human genetics : EJHG, 7(1), 20-26 (1999-03-27)
Coffin-Lowry syndrome (CLS) is an X-linked disorder characterized by facial dysmorphism, digit abnormalities and severe psychomotor retardation. CLS had previously been mapped to Xp22.2. Recently, mutations in the ribosomal S6 kinase (Rsk-2) gene were shown to be associated with CLS.
Andrew D Wingate et al.
The Biochemical journal, 393(Pt 3), 715-724 (2005-10-15)
Nur77 is a nuclear orphan receptor that is able to activate transcription independently of exogenous ligand, and has also been shown to promote apoptosis on its localization to mitochondria. Phosphorylation of Nur77 on Ser354 has been suggested to reduce ability

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service