Skip to Content
Merck
All Photos(8)

Key Documents

HPA003215

Sigma-Aldrich

Anti-OGFOD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-2-Oxoglutarate and iron-dependent oxygenase domain containing 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Check Cart for Availability


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EPENNQMAISNNSQQSNEQTDPEPEENETKKESSVPMCQGELRHWKTGHYTLIHDHSKAEFALDLILYCGCEGWEPEYGGFTSYIAKGEDEELLTVNPESNSLALVYRDRETLKFVKHINHRSLEQKKTFPNRTGFWDFSF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OGFOD1(55239)

General description

The gene OGFOD1 (2-oxoglutarate and iron-dependent oxygenase domain containing 1), also referred to as TPA1, is mapped to human chromosome 16q12.2. It is found to be overexpressed in breast cancer tissues.[1]

Immunogen

2-Oxoglutarate and iron-dependent oxygenase domain containing 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-OGFOD1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

OGFOD1 (2-oxoglutarate and iron-dependent oxygenase domain containing 1) gene encodes a member of the 2-oxoglutarate and iron dependent dioxygenase family that functions as a prolyl hydroxylase catalyzing the posttranslational hydroxylation of a highly conserved residue (Pro-62) in the small ribosomal protein S23 (RPS23). It plays the role of a translational regulator and its knockdown results in the formation of stress granules, translational arrest, and growth impairment. It functions in ischemic cell survival. It functions as a proapoptotic component by regulating the translation and HRI-mediated phosphorylation of eIF2α (eukaryotic translation initiation factor 2α) in cells recovering from arsenite-induced stress.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79904

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mathias Rask-Andersen et al.
Human genetics, 134(11-12), 1183-1193 (2015-09-06)
Single nucleotide polymorphisms (SNPs) within a genetic region including the first two introns of the gene encoding FTO have consistently been shown to be the strongest genetic factors influencing body mass index (BMI). However, this same also contains several regulatory
Rachelle S Singleton et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(11), 4031-4036 (2014-02-20)
2-Oxoglutarate (2OG) and Fe(II)-dependent oxygenase domain-containing protein 1 (OGFOD1) is predicted to be a conserved 2OG oxygenase, the catalytic domain of which is related to hypoxia-inducible factor prolyl hydroxylases. OGFOD1 homologs in yeast are implicated in diverse cellular functions ranging
Nahuel A Paolini et al.
American journal of human genetics, 100(3), 506-522 (2017-03-05)
Ribosomal protein (RP) gene mutations, mostly associated with inherited or acquired bone marrow failure, are believed to drive disease by slowing the rate of protein synthesis. Here de novo missense mutations in the RPS23 gene, which codes for uS12, are
Karen A Wehner et al.
Molecular and cellular biology, 30(8), 2006-2016 (2010-02-16)
Cells possess mechanisms that permit survival and recovery from stress, several of which regulate the phosphorylation of eukaryotic translation initiation factor 2alpha (eIF2alpha). We identified the human OGFOD1 protein as a novel stress granule component that regulates the phosphorylation of
Jae-Hwan Kim et al.
Oncotarget, 6(23), 19528-19541 (2015-04-25)
2-oxogluatrate and Fe(II)-dependent oxygenase domain-containing protein 1 (OGFOD1) was recently revealed to be a proline hydroxylase of RPS23 for translational termination. However, OGFOD1 is nuclear, whereas translational termination occurs in the cytoplasm, raising the possibility of another function of OGFOD1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service