Skip to Content
Merck
All Photos(7)

Key Documents

HPA002881

Sigma-Aldrich

Anti-S100A12 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CAAF1, Anti-CAGC, Anti-CGRP, Anti-Calcium-binding protein in amniotic fluid 1, Anti-Calgranulin-C, Anti-Neutrophil S100 protein, Anti-Protein S100-A12, Anti-S100 calcium-binding protein A12, Anti-p6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

KLEEHLEGIVNIFHQYSVRKGHFDTLSKGELKQLLTKELANTIKNIKDKAVIDEIFQGLDANQDEQVDFQEFISLVAIALKAAHYHTHKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100A12(6283)

General description

Protein S100-A12 is a protein encoded by the S100A12 gene in humans. The gene is composed of three exons, which are divided by two introns of 900bp and 400bp. The protein is encoded by both sequences in exons 2 and 3 (exon 2 codes for the N-terminal 45 amino acids and exon 3 codes for the C-terminal 46 amino acids).

Immunogen

Protein S100-A12 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Protein S100-A12 is an endogenous receptor ligand for advanced glycation end products. It acts as a neutrophil activation marker and plays a crucial role in pro-inflammatory cytokine. It also serves as a potential biomarker for a range of inflammatory diseases. In serum, it may be a potential biomarker for coronary plaque vulnerability with coronary artery disease (CAD). The gene induces vascular inflammation contributing to the development of atherosclerosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86241

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayssar A Elamin et al.
Frontiers in oncology, 9, 1484-1484 (2020-01-30)
Urothelial carcinoma of the urinary bladder (UCB) or bladder cancer remains a major health problem with high morbidity and mortality rates, especially in the western world. UCB is also associated with the highest cost per patient. In recent years numerous
Masaki Hara et al.
Nephron. Clinical practice, 123(3-4), 202-208 (2013-08-08)
S100A12 induces vascular inflammation contributing to the development of atherosclerosis. Serum S100A12 concentration is shown to be elevated in patients with chronic kidney disease (CKD), however the reason remains unclear. Transcriptional levels of S100A12 and RAGE (receptor for advanced glycation
R Wicki et al.
Cell calcium, 20(6), 459-464 (1996-12-01)
Here, we report the characterization of a human cDNA coding for the recently published amino acid sequence of a calcium-binding S100 protein, S100A12 (CGRP, calgranulin C, CAAF1, p6). The exon/intron structure of the S100A12 gene is similar to most other
Jun Liu et al.
Scandinavian journal of clinical and laboratory investigation, 74(2), 149-154 (2013-12-18)
The neutrophil activation marker S100A12 is an important pro-inflammatory cytokine and a potential biomarker for a range of inflammatory diseases. This study aims to investigate whether serum S100A12 concentrations are associated with angiographic coronary lesion complexity in patients with coronary
Yayoi Shiotsu et al.
BMC nephrology, 14, 16-16 (2013-01-18)
S100A12 protein is an endogenous receptor ligand for advanced glycation end products. In this study, the plasma S100A12 level was assessed as an independent predictor of mortality, and its utility in clinical settings was examined. In a previous cross-sectional study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service