Skip to Content
Merck
All Photos(3)

Key Documents

HPA002552

Sigma-Aldrich

Anti-FMNL3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Formin-like 3 isoform 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LLDVKELGRGMELIRRECSIHDNSVLRNFLSTNEGKLDKLQRDAKTAEEAYNAVVRYFGESPKTTPPSVFFPVFVRFIRSYKEAEQENEARKKQEEVMREKQLAQEAKKLDAKTPSQRNKWQQQE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FMNL3(91010)

General description

FMNL (formin like) encodes a formin homology protein. It belongs to the formin family with three other members: FMNL1, FMNL2 and FMNL3.

Immunogen

Formin-like 3 isoform 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

FMNL3 (formin like 3) is involved in the regulation of Rho-related signaling pathway by controlling the polarity, invasion, migration or metastasis. FMNL3 plays a major role in morphology regulation, migration and cytoskeletal organization. It can regulate endothelial cells (ECs) elongation during angiogenesis. During angiogenesis, activated FMNL3 stimulates the alignment of microtubules for remodeling of the EC cytoskeleton. FMNL3 also controls the protrusion of actin-rich filopodia structures that helps in enhanced migration and invasion.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86398

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Lynch et al.
PloS one, 8(11), e78428-e78428 (2013-11-14)
MiRNAs can have pleiotropic effects by targeting multiple genes belonging to diverse signalling networks. Alternatively, miRNAs can enhance the potency of their cellular effects by targeting multiple genes within the same genetic pathway. Previously, we and others have demonstrated that
Casey Eddington et al.
Journal of cell science, 137(4) (2024-01-24)
Filopodia are slender, actin-filled membrane projections used by various cell types for environment exploration. Analyzing filopodia often involves visualizing them using actin, filopodia tip or membrane markers. Due to the diversity of cell types that extend filopodia, from amoeboid to
Clare Hetheridge et al.
Journal of cell science, 125(Pt 6), 1420-1428 (2012-01-26)
The process of angiogenesis requires endothelial cells (ECs) to undergo profound changes in shape and polarity. Although this must involve remodelling of the EC cytoskeleton, little is known about this process or the proteins that control it. We used a
Masuko Katoh et al.
International journal of oncology, 22(5), 1161-1168 (2003-04-10)
FMNL (NM_005892.2) is a 5'-truncated partial cDNA encoding a Formin-homology protein related to DAAM1, DAAM2, DIAPH1 and DIAPH2. Here, we identified three members of FMNL gene family in the human genome by using bioinformatics. FMNL1 gene, corresponding to 5'-truncated KW-13
Siau Wei Bai et al.
BMC biology, 9, 54-54 (2011-08-13)
Cell migration is essential during development and in human disease progression including cancer. Most cell migration studies concentrate on known or predicted components of migration pathways. Here we use data from a genome-wide RNAi morphology screen in Drosophila melanogaster cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service