Skip to Content
Merck
All Photos(5)

Key Documents

HPA002550

Sigma-Aldrich

Anti-TTR antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Rabbit Anti-TTR, TTR Detection Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€482.00

€482.00


Check Cart for Availability


Select a Size

Change View
100 μL
€482.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

GTGESKCPLMVKVLDAVRGSPAINVAVHVFRKAADDTWEPFASGKTSESGELHGLTTEEEFVEGIYKVEIDTKSYWKALGISPFHEHAEVVFTANDSGPRRYTIAALLSPYSYSTTAVVTNPKE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TTR(7276)

General description

Transthyretin (TTR) was previously known as prealbumin. It is tetrameric protein with molecular mass of 55kDa. It consists of two distinct dimer-dimer interfaces which form two identical funnel-shaped T4 binding sites.

Immunogen

Transthyretin precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-TTR antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

TTR (transthyretin) protein is mainly involved in the plasma transport of both thyroxine and retinol (vitamin A) from bloodstream to brain. It is synthesized by the choroid plexus. It has characteristics of misfold and aggregate (amyloidogenesis) in vivo. Mutation in TTR gene is associated with the development of severe diseases such as familial amyloid neuropathy and Alzheimer′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77457

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I A Popov et al.
Molekuliarnaia biologiia, 47(6), 1011-1019 (2014-12-17)
Transthyretin, one of the major plasma proteins, has a number of posttranslational modifications and mutations, some of which are associated with the development of severe diseases, for instance, familial amyloid neuropathy and Alzheimer's disease. In order to investigate the role
Natalie J Galant et al.
Scientific reports, 6, 25080-25080 (2016-04-29)
Wild-type and mutant transthyretin (TTR) can misfold and deposit in the heart, peripheral nerves, and other sites causing amyloid disease. Pharmacological chaperones, Tafamidis(®) and diflunisal, inhibit TTR misfolding by stabilizing native tetrameric TTR; however, their minimal effective concentration is in
J Herbert et al.
Neurology, 36(7), 900-911 (1986-07-01)
Plasma transthyretin (TTR, formerly called prealbumin) is a 55-kd protein that participates in the plasma transport of both thyroxine and retinol (vitamin A). TTR concentrations are disproportionately high in human ventricular CSF, suggesting that TTR is either selectively transported across
Steven M Johnson et al.
Journal of molecular biology, 421(2-3), 185-203 (2012-01-17)
Transthyretin (TTR) is one of the many proteins that are known to misfold and aggregate (i.e., undergo amyloidogenesis) in vivo. The process of TTR amyloidogenesis causes nervous system and/or heart pathology. While several of these maladies are associated with mutations
T Liu et al.
Eye (London, England), 28(1), 26-33 (2013-10-12)
To describe the characteristic ophthalmic phenotypes of a large Chinese family with familial amyloid polyneuropathy due to a missense mutation in transthyretin (TTR) (c.307 C>G). Twenty-seven individuals (12 affected, 15 unaffected) from a five-generation Chinese family underwent general medical examination

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service