Skip to Content
Merck
All Photos(8)

Key Documents

HPA001907

Sigma-Aldrich

Anti-TERF2 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TTAGGG repeat-binding factor 2 antibody produced in rabbit, Anti-Telomeric DNA-binding protein antibody produced in rabbit, Anti-Telomeric repeat-binding factor 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Estimated to ship on12 April 2025



Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Estimated to ship on12 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

STGKEDKQPAPGPVEKPPREPARQLRNPPTTIGMMTLKAAFKTLSGAQDSEAAFAKLDQKDLVLPTQALPASPALKNKRPRKDENESSAPADGEGGSELQPKNKRMTISRLVLEEDSQSTEPS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TERF2(7014)

Looking for similar products? Visit Product Comparison Guide

General description

Telomeric binding protein, TERF2 (telomeric repeat binding factor 2), is also known as TRF2 and TRBF2 (TTAGGG repeat-binding factor 2), and is involved in protecting the guanine-rich, G-strand overhang, at the telomere terminus along with managing telomeric length. It has distant homology with TRF1 by possessing similar Myb-related DNA-binding motif. It is a component of shelterin complex.

Immunogen

Telomeric repeat-binding factor 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

By preventing the recognition of the telomeric end (5′-TTAGGG-3′ repeat) as double-strand breaks (DSBs), TERF2 (telomeric repeat binding factor 2) protects the ends. DSB response of TERF2 only depends on its basic domain but not Myb domain. This DSB response is independent of functional ataxia-telangiectasia mutated (ATM) and DNA-dependent protein kinases, MRE11 (double strand break repair nuclease)/Rad50 (double strand break repair protein)/NBS1 (Nijmegen breakage syndrome protein 1) complex and Ku70 (lupus Ku autoantigen protein p70), WRN (Werner syndrome RecQ like helicase) and BLM (Bloom syndrome RecQ like helicase) repair proteins.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86213

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan van Overbeek et al.
Current biology : CB, 16(13), 1295-1302 (2006-05-30)
Human chromosome ends are protected by shelterin, an abundant six-subunit protein complex that binds specifically to the telomeric-repeat sequences, regulates telomere length, and ensures that chromosome ends do not elicit a DNA-damage response (reviewed in). Using mass spectrometry of proteins
Paul S Bradshaw et al.
Nature genetics, 37(2), 193-197 (2005-01-25)
DNA damage surveillance networks in human cells can activate DNA repair, cell cycle checkpoints and apoptosis in response to fewer than four double-strand breaks (DSBs) per genome. These same networks tolerate telomeres, in part because the protein TRF2 prevents recognition
Lucile Delespaul et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(3), 857-867 (2016-08-17)
Despite various differences, nontranslocation-related sarcomas (e.g., comprising undifferentiated pleomorphic sarcoma, leiomyosarcoma, myxofibrosarcoma) are unified by their complex genetics. Extensive analysis of the tumor genome using molecular cytogenetic approaches showed many chromosomal gains, losses, and translocations per cell. Genomic quantitative alterations
D Broccoli et al.
Nature genetics, 17(2), 231-235 (1997-11-05)
Human telomeres are composed of long arrays of TTAGGG repeats that form a nucleoprotein complex required for the protection and replication of chromosome ends. One component of human telomeres is the TTAGGG repeat binding factor 1 (TRF1), a ubiquitously expressed
Åsa Ehlén et al.
Translational oncology, 4(4), 212-221 (2011-08-02)
The RNA-binding motif protein 3 (RBM3) was initially discovered as a putative cancer biomarker based on its differential expression in various cancer forms in the Human Protein Atlas (HPA). We previously reported an association between high expression of RBM3 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service