Skip to Content
Merck
All Photos(5)

Key Documents

HPA001864

Sigma-Aldrich

Anti-VAV1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Proto-oncogene vav

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LSWTPIAQNRGIMPFPTEEESVGDEDIYSGLSDQIDDTVEEDEDLYDCVENEEAEGDEIYEDLMRSEPVSMPPKMTEYDKRCCCLREIQQTEEKYTDTLGSIQQHFLKPLQRFLKPQDIEIIFINIEDLLRVHTHFLKEMKEAL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VAV1(7409)

General description

VAV1 (vav guanine nucleotide exchange factor 1), a 95kDa protein, is a guanosine diphosphate (GDP)-guanosine triphosphate (GTP) exchange factor of Rac. It is present in almost all hemopoietic cells with a Dbl homology domain, a hallmark of a guanine nucleotide exchange factor (GEF) for Rho-family GTPases.

Immunogen

Proto-oncogene vav recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VAV1 (vav guanine nucleotide exchange factor 1) helps to accelerate the exchange of bound GDP for GTP. The activity is controlled by phosphoinositide (PI) 3-kinase substrate through the Ras-dependent mechanism. VAV1 plays the core part in T cell antigen receptor (TCR)-dependent signaling pathways. Deficiency of VAV1 causes defective TCR-induced proliferation and cytokine synthesis in T-cells since VAV1 alters the activation signal of a calcium flux, extracellular signal-regulated kinase (ERK) and the nuclear factor κB (NF-κB) transcription factor. It has also been reported that in phospholipase Cγ1 (PLCγ1) activation, VAVA1 performs via both phosphoinositide-3-kinase (PI3K) dependent and independent pathways.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77489

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Matuszyk et al.
Postepy higieny i medycyny doswiadczalnej, 53(4), 531-543 (1999-11-02)
On the basis of recent reports we discuss the role of Vav in TCR-dependent signaling pathways. The Vav protein is GDP/GTP exchange factor for Rac, which initiates transduction of signals in JNK pathway. Upon stimulation of TCR by antigenic peptides
Yanting Shen et al.
Frontiers in oncology, 11, 761643-761643 (2021-11-23)
Existing prognostic risk assessment strategies for prostate cancer (PCa) remain unsatisfactory. Similar treatments for patients at the same disease stage can lead to different survival outcomes. Thus, we aimed to explore a novel immune landscape-based prognostic predictor and therapeutic target
Victor L J Tybulewicz et al.
Immunological reviews, 192, 42-52 (2003-04-03)
Vav1 is a 95-kDa protein expressed in all hemopoietic cells that becomes rapidly tyrosine phosphorylated following T cell antigen receptor (TCR) stimulation. Vav1 contains multiple domains characteristic of signal transducing proteins, including a Dbl homology domain, a hallmark of a
J Han et al.
Science (New York, N.Y.), 279(5350), 558-560 (1998-02-07)
Mitogen stimulation of cytoskeletal changes and c-jun amino-terminal kinases is mediated by Rac small guanine nucleotide-binding proteins. Vav, a guanosine diphosphate (GDP)-guanosine triphosphate (GTP) exchange factor for Rac that stimulates the exchange of bound GDP for GTP, bound to and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service